Skip to Content
Merck
All Photos(3)

Key Documents

P4902

Sigma-Aldrich

2-Phenylethyl β-D-thiogalactoside

≥98% (TLC)

Synonym(s):

PETG, Phenethyl β-D-thiogalactoside

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20O5S
CAS Number:
Molecular Weight:
300.37
Beilstein:
20033
MDL number:
UNSPSC Code:
12352201
PubChem Substance ID:
NACRES:
NA.25

Quality Level

Assay

≥98% (TLC)

form

powder

optical activity

[α]/D -33.00 to -27.00°, c = 9.00-11.00 mg/mL in methanol

technique(s)

thin layer chromatography (TLC): suitable

color

white

solubility

methanol: 49.00-51.00 mg/mL, clear, colorless

storage temp.

2-8°C

SMILES string

OC[C@H]1O[C@@H](SCCc2ccccc2)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C14H20O5S/c15-8-10-11(16)12(17)13(18)14(19-10)20-7-6-9-4-2-1-3-5-9/h1-5,10-18H,6-8H2/t10-,11+,12+,13-,14+/m1/s1

InChI key

ZNAMMSOYKPMPGC-HTOAHKCRSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Phenylethyl β-D-thiogalactoside is a cell-permable inhibitor of the reporter enzyme β-galactosidase.

Application

2-Phenylethyl β-D-thiogalactoside has been used in a study to assess subnanoliter enzymatic assays on microarrays. It has also been used in a study that investigated the integration of on-column immobilized enzyme reactor in microchip electrophoresis.

Other Notes

To gain a comprehensive understanding of our extensive range of Monosaccharides for your research, we encourage you to visit our Carbohydrates Category page.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pranav R H Joshi et al.
Molecular therapy. Methods & clinical development, 13, 279-289 (2019-03-20)
Despite numerous advancements in production protocols, manufacturing AAV to meet exceptionally high demand (1016-1017 viral genomes [VGs]) in late clinical stages and for eventual systemic delivery poses significant challenges. Here, we report an efficient, simple, scalable, robust AAV5 production process
Todd Galbraith et al.
Biotechnology journal, 14(1), e1800306-e1800306 (2018-11-30)
There is a strong clinical need to develop small-caliber tissue-engineered blood vessels for arterial bypass surgeries. Such substitutes can be engineered using the self-assembly approach in which cells produce their own extracellular matrix (ECM), creating a robust vessel without exogenous
Kyung-Han Lee et al.
European journal of nuclear medicine and molecular imaging, 31(3), 433-438 (2004-01-28)
There has recently been increasing interest in the development of radioprobes that specifically target proteins transcribed from expression of reporter genes of interest. The purpose of this study was to develop a radioprobe that targets one of the most widely
Philipp Angenendt et al.
Proteomics, 5(2), 420-425 (2005-02-09)
Many areas of research today are based on enzymatic assays most of which are still performed as enzyme-linked immunosorbent assays in microtiter plates. The demand for highly parallel screening of thousands of samples eventually led to a miniaturization and automation
R A Edwards et al.
Biochemistry, 29(49), 11001-11008 (1990-12-11)
The thermal denaturation of wild-type beta-galactosidase and two beta-galactosidases with substitutions at the active site was studied by kinetics, differential scanning calorimetry, electrophoresis, molecular exclusion chromatography, and circular dichroism. From the results, a model is developed for thermal denaturation of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service