Skip to Content
Merck
All Photos(3)

Documents

P3932

Sigma-Aldrich

Poly(2-hydroxyethyl methacrylate)

BioReagent, powder, suitable for cell culture

Synonym(s):

Poly(2-HEMA), Poly-HEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O3)n
CAS Number:
MDL number:
UNSPSC Code:
12352207
PubChem Substance ID:
NACRES:
NA.75

biological source

synthetic (organic)

product line

BioReagent

form

powder

technique(s)

cell culture | mammalian: suitable

solubility

ethanol: 120 mg/mL

density

1.15 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCO

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly(2-hydroxyethyl methacrylate) has been used:
  • as a nonadhesive substrate in the preparation of single cell suspensions
  • to implant into mice cornea for corneal micropocket assay
  • to coat the well plates for sphere assay using adipose-derived mesenchymal stem cells

Biochem/physiol Actions

Poly(2-hydroxyethyl methacrylate) is an acrylic compound and has dental usage. It is mainly used to inhibit cell adhesion to growth surfaces in culture vessels.

Physical form

Water-swellable polymer. Hydrogel.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

VAP-1-Mediated M2 Macrophage Infiltration Underlies IL-1beta-but Not VEGF-A-Induced Lymph-and Angiogenesis
Nakao S, et al.
The American Journal of Pathology, 178(4), 1913-1921 (2011)
Embryoid body formation from embryonic and induced pluripotent stem cells: Benefits of bioreactors
Rungarunlert S, et al.
World journal of stem cells, 1(1), 11-11 (2009)
Human cortical glial tumors contain neural stem-like cells expressing astroglial and neuronal markers in vitro
Ignatova Tatyana N, et al.
Glia, 39(3), 193-206 (2002)
Manuele Giuseppe Muraro et al.
Stem cells translational medicine, 1(8), 592-603 (2012-12-01)
Increasing evidence that cancers originate from small populations of so-called cancer stem cells (CSCs), capable of surviving conventional chemotherapies and regenerating the original tumor, urges the development of novel CSC-targeted treatments. Screening of new anticancer compounds is conventionally conducted on
Dario Barbone et al.
PloS one, 11(3), e0150044-e0150044 (2016-03-18)
To investigate the underlying causes of chemoresistance in malignant pleural mesothelioma, we have studied mesothelioma cell lines as 3D spheroids, which acquire increased chemoresistance compared to 2D monolayers. We asked whether the gene expression of 3D spheroids would reveal mechanisms

Protocols

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service