Skip to Content
Merck
All Photos(2)

Key Documents

192066

Sigma-Aldrich

Poly(2-hydroxyethyl methacrylate)

average Mv 300,000, crystalline

Synonym(s):

Poly(2-HEMA), Poly-HEMA

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(C6H10O3)n
CAS Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

form

crystalline

Quality Level

mol wt

average Mv 300,000

density

1.15 g/mL at 25 °C (lit.)

SMILES string

CC(=C)C(=O)OCCO

InChI

1S/C6H10O3/c1-5(2)6(8)9-4-3-7/h7H,1,3-4H2,2H3

InChI key

WOBHKFSMXKNTIM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Solid Poly(2-hydroxyethyl methacrylate) (pHEMA) is brittle, while pHEMA hydrogel is a soft material. Glass transition temperature for pHEMA hydrogels is reduced by increasing a content of water in the matrix. Macromolecular properties in pHEMA are characterized by non-covalent interactions of hydrogen-bonds among the polymer chains as well as the hydrated water molecules. The presence of polar groups of hydroxyl and carboxyl on each repeat unit makes this polymer compatible with water. The hydrophobic a-methyl groups of the backbone convey hydrolytic stability to the polymer and enhance mechanical strength of the polymer matrix.

Application

pHEMA scaffolds are used in tissue engineering. It can be used in hydrogels for biomedical applications, as sorbents for metal ions.pHEMA has been used in applications such as soft contact lenses, in drug delivery systems, and in kidney dialysis membranes.Nanoparticles of pHEMA may be employed as a carrier for controlled delivery of anticancer and antitumor drugs.

Physical form

Water-swellable polymer. Hydrogel.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

W E Roorda et al.
Pharmaceutical research, 5(11), 722-725 (1988-11-01)
The freezing and melting behavior of water in poly hydroxy ethyl methacrylate (pHEMA) hydrogels of different cross-linker and water contents was investigated in relation to the glass transition temperature (Tg) of the gels. After prolonged cooling at -15 degrees C
Fiber templating of poly (2-hydroxyethyl methacrylate) for neural tissue engineering
Flynn L,et al
Biomaterials, 24(23), 4265-4272 null
N Nogueira et al.
Journal of colloid and interface science, 385(1), 202-210 (2012-07-31)
The behavior of poly(2-hydroxyethyl methacrylate) (PHEMA) polymer monolayer spread on water was studied under various experimental conditions. The influence of subphase pH and temperature, compression speed, elapsed time from the deposit of the monolayer and the recording of the surface
Amit D Bhrany et al.
JAMA facial plastic surgery, 15(1), 29-33 (2013-01-19)
To evaluate the performance of a sphere-templated poly(2-hydroxyethyl methacrylate) (poly[HEMA]) tissue scaffold as a subcutaneous implant by comparing it with widely used high-density porous polyethylene (HDPPE) implant material. We implanted sphere-templated porous poly-(HEMA) and HDPPE disks into the dorsal subcutis
Yunfeng Li et al.
ACS applied materials & interfaces, 5(6), 2126-2132 (2013-02-23)
This paper presents a versatile way to prepare multiscale and gradient patterns of proteins. The protein patterns are fabricated by conjugating proteins covalently on patterns of polymer brush that are prepared by techniques combining colloidal lithography with photolithography, and two-step

Articles

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

Professor Shrike Zhang (Harvard Medical School, USA) discusses advances in 3D-bioprinted tissue models for in vitro drug testing, reviews bioink selections, and provides application examples of 3D bioprinting in tissue model biofabrication.

See All

Protocols

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Frequently asked questions (FAQs) for KAPA SYBR® FAST One-Step qRT-PCR Kits.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service