Skip to Content
Merck
All Photos(2)

Key Documents

P3501

Sigma-Aldrich

Phe-Gly-Gly

Synonym(s):

L-phenylalanylglycyl-glycine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H17N3O4
CAS Number:
Molecular Weight:
279.29
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.26

Assay

≥99%

Quality Level

form

powder

color

white

application(s)

cell analysis

storage temp.

−20°C

SMILES string

OC(CNC(CNC([C@@H](N)CC1=CC=CC=C1)=O)=O)=O

InChI

1S/C13H17N3O4/c14-10(6-9-4-2-1-3-5-9)13(20)16-7-11(17)15-8-12(18)19/h1-5,10H,6-8,14H2,(H,15,17)(H,16,20)(H,18,19)/t10-/m0/s1

InChI key

NAXPHWZXEXNDIW-JTQLQIEISA-N

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Structure and IR spectrum of phenylalanyl-glycyl-glycine tripetide in the gas-phase: IR/UV experiments, ab initio quantum chemical calculations, and molecular dynamic simulations.
Reha D, Valdes H, Vondrasek J, et al.
Chemistry (Easton), 11, 6803-6817 (2005)
S Fukayama et al.
Endocrinology, 126(4), 1941-1949 (1990-04-01)
We have investigated the actions of human PTH [hPTH-(1-34)] on the association of 45Ca2+ with two human (SaOS-2 and MG-63) and two rat (ROS 17/2.8 and UMR-106) osteoblast-like cell types. In SaOS-2 cells, hPTH-(1-34) binds to specific membrane receptors to
Aus T Ali et al.
Annals of clinical biochemistry, 43(Pt 3), 207-213 (2006-05-18)
A previous study has demonstrated that alkaline phosphatase (AP) may play a role in the control of intracellular lipid accumulation in the rodent preadipocyte cell line, 3T3-L1. The present study investigated whether AP may have a similar function in preadipocytes
Pingxiu Tang et al.
Talanta, 178, 650-655 (2017-11-16)
Hypercrosslinked polymers (HCPs) are currently receiving great attention due to their unique characteristics and potential uses in diverse areas. However, the field of HCPs for open-tubular capillary electrochromatography (OT-CEC) separations has not been explored. Here, a knitted aromatic polymer (KAP)
Antonio Nebbioso et al.
Rapid communications in mass spectrometry : RCM, 24(21), 3163-3170 (2010-10-14)
A model heterogeneous mixture of a hydrophilic tripeptide (phenylalanine-glycine-glycine, PGG) and hydrophobic organic acids ((12)C- and (13)C-octanoic acid and pentadecanoic acid) was subjected to electrospray ionization mass spectrometry (ESI-MS). The objective was to verify the previously noted inconsistencies in ESI-MS

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service