Skip to Content
Merck
All Photos(4)

Key Documents

P2533

Sigma-Aldrich

Poly-L-ornithine hydrochloride

suitable for ligand binding assays, Mol wt 15,000-30,000

Synonym(s):

L-Ornithine homopolymer hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.26

Product Name

Poly-L-ornithine hydrochloride, mol wt 15,000-30,000

form

powder or solid

Quality Level

mol wt

15,000-30,000

technique(s)

ligand binding assay: suitable

color

white to off-white

storage temp.

−20°C

SMILES string

Cl.NCCCC(N)C(O)=O

InChI key

GGTYBZJRPHEQDG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Poly-L-ornithine hydrochloride has been used as a substrate analog of acetyl-poly-L-lysine to study substrate binding and catalysis of the NAD-dependent protein deacetylase Hst2. It has been used to bind alginate-polycation-alginate (APA) microcapsules.

Biochem/physiol Actions

Poly-L-ornithine is a polycationic synthetic amino acid polymer. It boosts nasal absorption of hydrophilic macromolecular drugs. Poly-L-ornithine is a specialized coating that acts as a charge modifier.

Analysis Note

Molecular weight based on viscosity.

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hierarchical Structure Formation of Cylindrical Brush Polymer?Surfactant Complexes
Cong, Y. et al.
Langmuir, 25, 6392-6397 (2005)
Kun Luo et al.
Journal of integrative neuroscience, 20(3), 529-539 (2021-10-15)
Rab3a, a subtype protein in the Rab3 family amongst the small G proteins, is closely associated with the learning and memory formation process. Various neuronal stimuli can induce the expression of Rab3a; however, how DNA modification is involved in regulating
Preparation and Evaluation of PEGylated Poly-L-ornithine Complex as a Novel Absorption Enhancer
Yusuke K
Biologicals : Journal of the International Association of Biological Standardization (2017)
Use of Substrate Analogs and Mutagenesis to Study Substrate Binding and Catalysis in the Sir2 Family of NAD-dependent Protein Deacetylases*
Ahlia N
The Journal of Biological Chemistry (2006)
Emmanuel C. Opara
Methods in Molecular Biology (2017)

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service