Skip to Content
AE
EN
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Back
Ponceau Xylidine
Synonym(s):
Acid Red 26, Ponceau 2 R, Xylidine ponceau, Xylidine ponceau 2R
Empirical Formula (Hill Notation):
C
18
H
14
N
2
Na
2
O
7
S
2
CAS Number:
3761-53-3
Molecular Weight:
480.42
EC Number:
223-178-3
Products
Product Category
stains (1)
Brand
Sigma-Aldrich (1)
Supelco (1)
Formula Weight
Physical Form
Purity
Format
Chemical Composition
Quality Segment
Application
Reference Material Type
Special Grade
analytical standard (1)
Technique
Filter Products
Showing 1-2 of 2
Compare
Product Number
Description
Pricing
P2395
Ponceau Xylidine,
Dye content ≥60 %, Powder
Expand
View Pricing
22308
Ponceau Xylidine,
analytical standard
Expand
View Pricing