Skip to Content
Merck
All Photos(1)

Key Documents

P215

Sigma-Aldrich

PD 98,059

solid

Synonym(s):

2-(2-Amino-3-methoxyphenyl)-4H-1-benzopyran-4-one, PD98059

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H13NO3
CAS Number:
Molecular Weight:
267.28
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

form

solid

color

yellow

solubility

DMSO: 10 mg/mL, clear

storage temp.

−20°C

SMILES string

COc1cccc(c1N)C2=CC(=O)c3ccccc3O2

InChI

1S/C16H13NO3/c1-19-14-8-4-6-11(16(14)17)15-9-12(18)10-5-2-3-7-13(10)20-15/h2-9H,17H2,1H3

InChI key

QFWCYNPOPKQOKV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

PD 98,059 has been used:
  • in growth medium as a mitogen-activated protein (MAP) kinase inhibitor, in order to study the secondary messengers involved in the effect of D1-like receptors
  • in calcium-free RPMI (Roswell park memorial institute)-1640 medium to study its effect on inflammation, proliferation and differentiation in human epidermal keratinocyte cell line in conjunction of cystic fibrosis transmembrane conductance regulator (CFTR) knockdown
  • in culture medium for cell migration assay

Biochem/physiol Actions

PD 98,059 is a flavonoid and specific inhibitor of mitogen-activated protein kinase kinase (MAPKK). In mice, PD 98,059 helps to block zymosan stimulated organ dysfunction syndrome and non-septic shock. It is known to inhibit in vitro hypertrophy. PD 98,059 also induces cartilage formation in mesenchymal stromal cells.

Features and Benefits

This compound is featured on the MAPKs page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Legal Information

Sold with permission of Warner Lambert Company.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Sepsis: Advances in Research and Treatment: 2011 Edition, 38-38 (2012)
Epidermal CFTR suppresses MAPK/NF-kappaB to promote cutaneous wound healing
Chen J
Cellular Physiology and Biochemistry, 39(6), 2262-2274 (2016)
Lingling Zhang et al.
European journal of pharmacology, 792, 15-25 (2016-11-05)
Abnormal proliferation and hypertrophy of vascular smooth muscle (VSMC), as the main structural component of the vasculature, is an important pathological mechanism of hypertension. Recently, increased levels of arginine vasopressin (AVP) and copeptin, the C-terminal fragment of provasopressin, have been
D1-like receptors inhibit insulin-induced vascular smooth muscle cell proliferation via down-regulation of insulin receptor expression
Zeng C, et al.
Journal of Hypertension, 27(5), 1033-1033 (2009)
Ki-Sun Park et al.
Nucleic acids research, 45(22), 12766-12779 (2017-12-16)
Imprinted genes occur in discrete clusters that are coordinately regulated by shared DNA elements called Imprinting Control Regions. H19 and Igf2 are linked imprinted genes that play critical roles in development. Loss of imprinting (LOI) at the IGF2/H19 locus on

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service