Skip to Content
Merck
All Photos(1)

Key Documents

559389

Sigma-Aldrich

SB 203580

≥98% (HPLC), film (Thin), p38 MAP kinase inhibitor, Calbiochem®

Synonym(s):

SB 203580, 4-(4-Fluorophenyl)-2-(4-methylsulfinylphenyl)-5-(4-pyridyl)1H-imidazole, p38 MAP Kinase Inhibitor XVI

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H16FN3OS
CAS Number:
Molecular Weight:
377.43
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Product Name

SB 203580, SB 203580, CAS 152121-47-6, is a highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC₅₀ = 34 nM in vitro, 600 nM in cells).

Quality Level

Assay

≥98% (HPLC)

form

film (Thin)

manufacturer/tradename

Calbiochem®

storage condition

OK to freeze
protect from light

solubility

DMSO: 30 mg/mL

shipped in

ambient

storage temp.

−20°C

SMILES string

Fc1ccc(cc1)c2nc([nH]c2c4ccncc4)c3ccc(cc3)[S+]([O-])C

InChI

1S/C21H16FN3OS/c1-27(26)18-8-4-16(5-9-18)21-24-19(14-2-6-17(22)7-3-14)20(25-21)15-10-12-23-13-11-15/h2-13H,1H3,(H,24,25)

InChI key

CDMGBJANTYXAIV-UHFFFAOYSA-N

General description

Reduces epirubicin-induced cell injury and caspase-3/7 activity. A highly specific, potent, cell-permeable, selective, reversible, and ATP-competitive inhibitor of p38 MAP kinase (IC50 = 34 nM in vitro, 600 nM in cells). Does not significantly inhibit the JNK and p42 MAP kinase at 100 µM. Inhibits IL-1 and TNF-α production from LPS-stimulated human monocytes and the human monocyte cell line THP-1 (IC50 = 50-100 nM). Inhibits bone morphogenetic protein-2-induced neurite outgrowth in PC12 cells. Also inhibits platelet aggregation caused by collagen (IC50 = 0.2-1.0 µM) or the thromboxane analog U-46619 (Cat. No. 538944). A 1 mg/ml solution of SB 203580 (Cat. No. 559398) in anhydrous DMSO is also available.

Biochem/physiol Actions

Cell permeable: yes
Product competes with ATP.
Reversible: yes
Target IC50: 34 nM against p38 MAP kinase in vitro, 600 nM in cells; 50-100 nM against IL-1 and TNF-α production from LPS-stimulated human monocytes and the human monocyte cell line THP-1; 0.2-1.0 µM against platelet aggregation caused by collagen

Warning

Toxicity: Irritant (B)

Other Notes

Yamada. T., et al. 2012. Free Radic. Biol. Med. (In press)
Powell, D.J., et al. 2003. Mol. Cell Biol.23, 7794.
Davies, S.P., et al. 2000. Biochem. J.351, 95.
Iwasaki, S., et al. 1999. J. Biol. Chem.274, 26503.
Gallagher, T.F., et al. 1997. Bioorg. Med. Chem. 5, 49.
LoGrasso, P.V., et al. 1997. Biochem. 36, 10422.
Hazzalin, C.A., et al. 1996. Curr. Biol.6, 1028.
Kramer, R.M., et al. 1996. J. Biol. Chem.271, 27723.
Saklatvala, J., et al. 1996. J. Biol. Chem. 271, 6586.
Cuenda, A., et al. 1995. FEBS Lett.364, 229.
Gallagher, T.F., et al. 1995. Bioorg. Med. Chem. Lett. 5, 1171.
Lee, J.C., et al. 1994. Nature 372, 739.

Legal Information

CALBIOCHEM is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Suji Bae et al.
Molecules (Basel, Switzerland), 26(6) (2021-04-04)
Matrix metalloproteinases (MMPs), key molecules of cancer invasion and metastasis, degrade the extracellular matrix and cell-cell adhesion molecules. MMP-10 plays a crucial role in Helicobacter pylori-induced cell-invasion. The mitogen-activated protein kinase (MAPK) signaling pathway, which activates activator protein-1 (AP-1), is
Matthew Herrera et al.
American journal of physiology. Cell physiology, 321(1), C104-C116 (2021-04-29)
Chronic alcohol alters the immune system enhancing the susceptibility to inflammation, bacterial, and viral infections in alcohol users. We have shown that alcohol causes increased permeability of mesenteric lymphatic vessels in alcohol-fed rats. The mechanisms of alcohol-induced lymphatic leakage are
John R Ortaldo et al.
Journal of immunology (Baltimore, Md. : 1950), 170(4), 1763-1769 (2003-02-08)
NK cells can express both activating and inhibitory Ly49 receptors on their cell surface. When cells expressing both receptors are presented with a ligand, inhibition dominates the functional outcome. In this report we demonstrate that costimulation of the activating Ly49D
Shihab Kochumon et al.
Biomolecules, 11(9) (2021-09-29)
IP-10 (also called CXCL10) plays a significant role in leukocyte homing to inflamed tissues, and increased IP-10 levels are associated with the pathologies of various inflammatory disorders, including type 2 diabetes, atherosclerosis, and cancer. TNF-α is a potent activator of
Milica Bozic et al.
Nature communications, 11(1), 1943-1943 (2020-04-25)
Kidney fibrosis is a highly deleterious process and a final manifestation of chronic kidney disease. Alpha-(α)-synuclein (SNCA) is an actin-binding neuronal protein with various functions within the brain; however, its role in other tissues is unknown. Here, we describe the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service