Skip to Content
Merck
All Photos(1)

Key Documents

P1259

Sigma-Aldrich

Phytase from wheat

≥0.01 unit/mg solid

Synonym(s):

6-Phytase, Phytate 6-phosphatase, myo-Inositol-hexakisphosphate 6-phosphohydrolase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

powder

specific activity

≥0.01 unit/mg solid

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Phytase catalyzes the hydrolysis of phytic acid to inositol and free orthophosphate.

Application

Phytase has been used in a study to quantify the number of feedstuffs showing significant phytase activity. It has also been used in a study to investigate the effect of adjusting the pH to 5.0 (optimum for wheat phytase) during whole wheat bread making on phytase activity.

Biochem/physiol Actions

Wheat phytase can degrade inositol phosphate-6 and 5 at a pH of 4 and 5.

Quality

Crude

Unit Definition

One unit will liberate 1.0 μmole of inorganic phosphorus from 1.5 mM phytate per min at pH 5.15 at 55 °C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chandra Shekar Mootapally et al.
Journal of molecular microbiology and biotechnology, 26(4), 252-260 (2016-05-14)
Phytases have been widely used as animal feed supplements to increase the availability of digestible phosphorus, especially in monogastric animals fed cereal grains. The present study describes the identification of a full-length phytase gene of Prevotella species present in Mehsani
Elsa Arcalis et al.
Plant molecular biology, 83(1-2), 105-117 (2013-04-05)
Many plant-based systems have been developed as bioreactors to produce recombinant proteins. The choice of system for large-scale production depends on its intrinsic expression efficiency and its propensity for scale-up, post-harvest storage and downstream processing. Factors that must be considered
Total phosphorus, phytate-phosphorus and phytase activity in plant feedstuffs
Eeckhout, W. and M. Paepe
Anim. Feed Sci. Technol., 47, 19-29 (1994)
J C González-Vega et al.
Journal of animal science, 91(10), 4807-4816 (2013-08-15)
An experiment was conducted to test the hypothesis that values for apparent total tract digestibility (ATTD) of Ca in pigs are influenced by endogenous Ca lost from the gastrointestinal tract. The objective was to determine the endogenous loss of Ca
Katrine Pontoppidan et al.
Archives of animal nutrition, 66(6), 431-444 (2012-10-27)
Phytases (EC 3.1.3) are widely used in animal feed to increase the availability of phosphorus and decrease the anti nutritive effect of myo-inositol hexakisphosphate (InsP₆). The aim of this work was to investigate the stereospecific degradation of InsP₆ in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service