Skip to Content
Merck
All Photos(2)

Key Documents

O9504

Sigma-Aldrich

Oxaloacetic acid

powder, suitable for hybridoma

Synonym(s):

2-Oxosuccinic acid, Ketosuccinic acid, Oxalacetic acid, Oxobutanedioic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HOOCCH2COCOOH
CAS Number:
Molecular Weight:
132.07
Beilstein:
1705475
EC Number:
MDL number:
UNSPSC Code:
12352205
PubChem Substance ID:
NACRES:
NA.75

Assay

≥97% (HPLC)

form

powder

technique(s)

cell culture | hybridoma: suitable

impurities

endotoxin, tested

solubility

H2O: 100 mg/mL

storage temp.

−20°C

SMILES string

OC(=O)CC(=O)C(O)=O

InChI

1S/C4H4O5/c5-2(4(8)9)1-3(6)7/h1H2,(H,6,7)(H,8,9)

InChI key

KHPXUQMNIQBQEV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Use as a TCA (Krebs cycle) intermediate supplement in hybridoma cell culture applications. Enhances hybridoma growth and productivity.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lee Hua Long et al.
Biochemical and biophysical research communications, 417(1), 446-450 (2011-12-15)
Several phenolic compounds as well as ascorbate can oxidise in certain cell culture media (especially Dulbecco's modified Eagle's medium (DMEM)) to generate hydrogen peroxide. Addition of oxaloacetate decreased the levels of H(2)O(2) detected and the oxaloacetate was depleted. Oxaloacetate was
Tonya N Zeczycki et al.
Biochemistry, 50(45), 9724-9737 (2011-10-01)
The catalytic mechanism of the MgATP-dependent carboxylation of biotin in the biotin carboxylase domain of pyruvate carboxylase from R. etli (RePC) is common to the biotin-dependent carboxylases. The current site-directed mutagenesis study has clarified the catalytic functions of several residues
L T Wong et al.
Pediatric research, 20(3), 274-279 (1986-03-01)
An infant with the acute neonatal form of pyruvate carboxylase deficiency (cross-reacting material negative) presented with severe intractable lactic acidosis within 4 h after birth. He also had hyperammonemia, hypercitrullinemia, and hyperlysinemia. Plasma glutamine was not elevated. He had a
Alexander Zlotnik et al.
Anesthesiology, 116(1), 73-83 (2011-12-02)
Decreasing blood glutamate concentrations after traumatic brain injury accelerates brain-to-blood glutamate efflux, leading to improved neurologic outcomes. The authors hypothesize that treatment with blood glutamate scavengers should reduce neuronal cell loss, whereas administration of glutamate should worsen outcomes. The authors
Randy J Giedt et al.
Free radical biology & medicine, 52(2), 348-356 (2011-11-22)
Ischemia (I)/reperfusion (RP)-induced endothelial cell (EC) injury is thought to be due to mitochondrial reactive oxygen species (mtROS) production. MtROS have been implicated in mitochondrial fission. We determined whether cultured EC exposure to simulated I/RP causes morphological changes in the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service