Skip to Content
Merck
All Photos(1)

Key Documents

N8912

Sigma-Aldrich

Monoclonal Anti-NG2 antibody produced in mouse

clone 132.38, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-Chondroitin Sulfate Proteoglycan Melanoma-associated, Anti-MCSP, Anti-MCSPG, Anti-MEL-CSPG MSK16 Chondroitin Sulfate Proteoglycan NG2

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

132.38, monoclonal

form

buffered aqueous solution

species reactivity

rat

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
indirect ELISA: suitable
microarray: suitable
western blot: 1 μg/mL using rat brain neonatal extract

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

General description

Chondroitin sulfate proteoglycan 4 (CSPG4) or nerve/glial antigen 2 (NG2) is a growth-inhibitory chondroitin sulfate proteoglycan (CSPG) produced after spinal cord injury. It has a single glycosaminoglycan (GAG) chain. The protein belongs to the transmembrane chondroitin sulfate proteoglycans family.

Immunogen

cell extract of HEK293 cells expressing the D3 domain of rat NG2.

Application

Applications in which this antibody has been used successfully, and the associated peer-reviewed papers, are given below.
Immunohistochemistry (1 paper)
Western Blotting

Biochem/physiol Actions

Chondroitin sulfate proteoglycan 4 (CSPG4) or nerve/glial antigen 2 (NG2) stimulates the activation of integrins in dorsal root ganglion neurons. It also has a role in stimulating inflammatory cytokine expression in microglia.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NG2, a member of chondroitin sulfate proteoglycans family mediates the inflammatory response of activated microglia.
Gao Q, ey .al
Neuroscience and Behavioral Physiology (2010)
Entrapment via Synaptic-Like Connections between NG2 Proteoglycan+ Cells and Dystrophic Axons in the Lesion Plays a Role in Regeneration Failure after Spinal Cord Injury
Angela R
The Journal of Neuroscience (2014)
Stefano Di Santo et al.
PloS one, 4(5), e5643-e5643 (2009-05-30)
Current evidence suggests that endothelial progenitor cells (EPC) contribute to ischemic tissue repair by both secretion of paracrine factors and incorporation into developing vessels. We tested the hypothesis that cell-free administration of paracrine factors secreted by cultured EPC may achieve
Antibody-based proteomics for esophageal cancer: Identification of proteins in the nuclear factor-?B pathway and mitotic checkpoint
Norihisa U
Cancer Science (2009)
Genetically Engineered Artificial Microvesicles Carrying Nerve Growth Factor Restrains the Progression of Autoimmune Encephalomyelitis in an Experimental Mouse Model.
Alatrash, et al.
International Journal of Molecular Sciences, 24 (2023)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service