Skip to Content
Merck
All Photos(1)

Key Documents

N8133

Sigma-Aldrich

Nerve Growth Factor from Vipera lebetina venom

lyophilized powder, suitable for cell culture

Synonym(s):

NGF

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.77

Product Name

Nerve Growth Factor from Vipera lebetina venom, NGF, lyophilized powder, suitable for cell culture

biological source

Vipera Lebetina

Quality Level

form

lyophilized powder

potency

0.2-20 μg/mL ED50/EC50

quality

endotoxin tested

mol wt

protein 32.5 kDa

packaging

pkg of 0.1 mg

technique(s)

cell culture | mammalian: suitable

impurities

<10 EU/vial

color

white to off-white

UniProt accession no.

storage temp.

−20°C

Physical form

Lyophilized from a 0.2 μm filtered solution in sodium acetate buffer.

Preparation Note

Isolated from the snake venom of V. lebetina.

Analysis Note

The biological activity of Nerve Growth Factor (NGF) from Vipera lebetina venom is measured in a cell proliferation assay using PC-12 cells.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Giusi Prencipe et al.
Journal of immunology (Baltimore, Md. : 1950), 192(7), 3345-3354 (2014-03-04)
Nerve growth factor (NGF) levels are highly increased in inflamed tissues, but their role is unclear. We show that NGF is part of a regulatory loop in monocytes: inflammatory stimuli, while activating a proinflammatory response through TLRs, upregulate the expression
Lucy Hepburn et al.
Science (New York, N.Y.), 346(6209), 641-646 (2014-11-02)
Many key components of innate immunity to infection are shared between Drosophila and humans. However, the fly Toll ligand Spaetzle is not thought to have a vertebrate equivalent. We have found that the structurally related cystine-knot protein, nerve growth factor
Jeremy E Purvis et al.
Cell, 152(5), 945-956 (2013-03-05)
A growing number of studies are revealing that cells can send and receive information by controlling the temporal behavior (dynamics) of their signaling molecules. In this Review, we discuss what is known about the dynamics of various signaling networks and
M Florencia Iulita et al.
Trends in pharmacological sciences, 35(7), 338-348 (2014-06-26)
Alzheimer's disease (AD) is a devastating neurodegenerative condition and the most common type of amnestic dementia in the elderly. Individuals with Down syndrome (DS) are at increased risk of developing AD in adulthood as a result of chromosome 21 trisomy
Geraldine Longo et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 33(24), 10066-10074 (2013-06-14)
Although chronic pain is the most common symptom of arthritis, relatively little is known about the mechanisms driving it. Recently, a sprouting of autonomic sympathetic fibers into the upper dermis of the skin, an area that is normally devoid of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service