Skip to Content
Merck
All Photos(1)

Documents

N7410

Sigma-Aldrich

β-Nicotinamide adenine dinucleotide, reduced disodium salt

~98%, pkg of 2 mg (per vial)

Synonym(s):

β-DPNH, β-NADH, Coenzyme I reduced disodium salt, Diphosphopyridine nucleotide reduced disodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N7Na2O14P2
CAS Number:
Molecular Weight:
709.40
Beilstein:
5230241
EC Number:
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.51

Assay

~98%

Quality Level

form

solid

packaging

pkg of 2 mg (per vial)

storage temp.

room temp

SMILES string

[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@H]2O[C@@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]3O[C@H]([C@H](O)[C@@H]3O)n4cnc5c(N)ncnc45)[C@H](O)[C@@H]2O

InChI

1S/C21H29N7O14P2.2Na/c22-17-12-19(25-7-24-17)28(8-26-12)21-16(32)14(30)11(41-21)6-39-44(36,37)42-43(34,35)38-5-10-13(29)15(31)20(40-10)27-3-1-2-9(4-27)18(23)33;;/h1,3-4,7-8,10-11,13-16,20-21,29-32H,2,5-6H2,(H2,23,33)(H,34,35)(H,36,37)(H2,22,24,25);;/q;2*+1/p-2/t10-,11+,13-,14+,15-,16+,20-,21+;;/m0../s1

InChI key

QRGNQKGQENGQSE-QUWMEQBESA-L

Looking for similar products? Visit Product Comparison Guide

Application

β-Nicotinamide adenine dinucleotide (NAD+) and β-Nicotinamide adenine dinucleotide, reduced (NADH) comprise a coenzyme redox pair (NAD+:NADH) involved in a wide range of enzyme catalyzed oxidation reduction reactions. In addition to its redox function, NAD+/NADH is a donor of ADP-ribose units in ADP-ribosylaton (ADP-ribosyltransferases; poly(ADP-ribose) polymerases ) reactions and a precursor of cyclic ADP-ribose (ADP-ribosyl cyclases).

Biochem/physiol Actions

Electron donor

Reconstitution

Solutions should be prepared fresh and used promptly.

Other Notes

Packaged based on NADH content as determined by UV-Vis.
This is the common form of NADH; do not confuse with α-NADH.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 2

1 of 2

Qiuju Ding et al.
Life (Basel, Switzerland), 10(9) (2020-09-26)
With the advent of next generation sequencing, the list of mitochondrial DNA (mtDNA) mutations identified in patients rapidly and continuously expands. They are frequently found in a limited number of cases, sometimes a single individual (as with the case herein
Amy Swanston et al.
Nucleic acids research, 47(12), 6172-6183 (2019-05-21)
Topoisomerase II (Top2) is an essential enzyme that decatenates DNA via a transient Top2-DNA covalent intermediate. This intermediate can be stabilized by a class of drugs termed Top2 poisons, resulting in massive DNA damage. Thus, Top2 activity is a double-edged
Keith J Mickolajczyk et al.
Proceedings of the National Academy of Sciences of the United States of America, 117(31), 18459-18469 (2020-07-23)
Mdn1 is an essential mechanoenzyme that uses the energy from ATP hydrolysis to physically reshape and remodel, and thus mature, the 60S subunit of the ribosome. This massive (>500 kDa) protein has an N-terminal AAA (ATPase associated with diverse cellular
Maali Odeh et al.
The FEBS journal, 287(1), 73-93 (2019-09-24)
Physiological or pathological muscle disuse/inactivity or loss of the neural-muscular junction cause muscle atrophy. Atrophy-inducing conditions cause metabolic oxidative stress in the muscle tissue, activation of the ubiquitin-proteasome and of the autophagosome-lysosome systems, enhanced removal of the damaged proteins and
Richard P Ebstein et al.
FEBS letters, 585(11), 1529-1536 (2011-05-12)
Increasing evidence suggests that the nonapeptide, oxytocin (OT), helps shape social and affiliative behaviors not only in lower mammals but also in humans. Recently, an essential mediator of brain OT release has been discovered, ADP-ribosyl cyclase and/or CD38. We have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service