Merck
  • Are retinoids potential therapeutic agents in disorders of social cognition including autism?
Are retinoids potential therapeutic agents in disorders of social cognition including autism?
FEBS letters (2011-05-12)
Richard P Ebstein, David Mankuta, Nurit Yirmiya, Fabio Malavasi
ABSTRACT
Increasing evidence suggests that the nonapeptide, oxytocin (OT), helps shape social and affiliative behaviors not only in lower mammals but also in humans. Recently, an essential mediator of brain OT release has been discovered, ADP-ribosyl cyclase and/or CD38. We have subsequently shown that polymorphisms across the CD38 gene are associated with autism spectrum disorders (ASD). Notably, CD38 expression in lymphoblastoid cells (LBC) is reduced in cell lines derived from ASD subjects compared to parental cell lines. Intriguingly, a correlation was observed between CD38 expression and measures of social function in ASD. Finally, we have shown that all-trans retinoic acid (ATRA), a known inducer of CD38 transcription, can rescue low CD38 expressing LBC lines derived from ASD subjects and restore normal levels of transcription of this ectoenzyme providing 'proof of principle' in a peripheral model that retinoids are potential therapeutic agents in ASD.
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ~98%, pkg of 100 mg
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ≥94% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt hydrate, ≥97% (HPLC)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 0.2 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 2 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 1 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 5 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced disodium salt, ~98%, pkg of 25-30 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, reduced dipotassium salt
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide lithium salt from Saccharomyces cerevisiae, ≥95%