Skip to Content
Merck
All Photos(2)

Documents

N4288

Sigma-Aldrich

Anti-NAV3 (C-terminal) antibody produced in rabbit

enhanced validation

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-Neuron navigator 3, Anti-Pore membrane and/or filament interacting like protein 1 (POMFIL1), Anti-STEERIN3, Anti-human UNC-53 homolog (unc53H3)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen ~280 kDa

species reactivity

human, mouse (predicted), rat (predicted)

packaging

antibody small pack of 25 μL

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

concentration

~1 mg/mL

technique(s)

western blot: 1-2 μg/mL using HEK-292T cells lysate expressing human NAV3 fusion protein

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NAV3(89795)
mouse ... Nav3(260315)
rat ... Nav3(314814)

General description

NAV3 belongs to the neuron navigator (NAV) family of proteins. NAV3 is expressed in adult heart, kidney, and brain.
Neuron navigator 3 (NAV3) is highly expressed in tumors. It possesses actin-binding domains and the gene encoding it is localized on chromosome 12q21.

Application

Anti-NAV3 (C-terminal) antibody has been used in immunoblotting, western blotting.

Biochem/physiol Actions

NAV3 has been suggested to function as a novel tumor suppressor gene. Deletions and translocation affecting NAV3 have been shown to occur in patients with advanced mycosis fungoides or Sézary syndrome, the most common forms of primary cutaneous T-cell lymphoma (CTCL).
Neuron navigator 3 (NAV3) has been shown to control migration and invasion of epithelial cells. Once it is activated by growth factors, NAV3 occupies the ends of microtubules and stimulates their growth.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Pilvi Maliniemi et al.
Experimental dermatology, 20(11), 926-931 (2011-10-15)
The neuron navigator 3 (NAV3) gene on chromosome 12q21 encodes a microtubule plus end tracking protein and belongs to the navigator family of cytoskeletal regulators. Loss of heterozygosity on 12q has previously been suggested to be associated with poor prognosis
Jasmine M Aly et al.
Reproductive sciences (Thousand Oaks, Calif.), 27(3), 925-934 (2020-02-13)
NAV 3 is a tumor suppressor of unknown function in leiomyomas. The objective of this study is to assess NAV3 expression and its potential role in human uterine leiomyomas. NAV3 protein expression was examined in patient leiomyoma and patient-matched myometrial
Primary cutaneous T-cell lymphomas show a deletion or translocation affecting NAV3, the human UNC-53 homologue
Karenko Leena, et al.
Cancer Research, 65(18), 8101-8110 (2005)
Hadas Cohen-Dvashi et al.
EMBO molecular medicine, 7(3), 299-314 (2015-02-14)
Dissemination of primary tumor cells depends on migratory and invasive attributes. Here, we identify Navigator-3 (NAV3), a gene frequently mutated or deleted in human tumors, as a regulator of epithelial migration and invasion. Following induction by growth factors, NAV3 localizes
Ryan Charles Pink et al.
Gynecologic oncology, 137(1), 143-151 (2015-01-13)
Ovarian cancer is the deadliest gynaecological cancer. A major contributor to the poor survival rate is the development of chemoresistance to platinum-based therapies such as cisplatin and carboplatin. Here we aimed to test the role of miRNAs in the acquisition

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service