Skip to Content
Merck
All Photos(3)

Documents

N3378

Sigma-Aldrich

4-Nitrophenyl β-D-fuco­pyran­oside

≥98% (TLC), powder

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H15NO7
CAS Number:
Molecular Weight:
285.25
Beilstein:
1291394
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

product name

4-Nitrophenyl β-D-fuco­pyran­oside, ≥98% (TLC)

Assay

≥98% (TLC)

form

powder

solubility

ethanol: soluble 25 mg/mL, clear, colorless to faintly yellow

storage temp.

−20°C

SMILES string

C[C@H]1O[C@@H](Oc2ccc(cc2)[N+]([O-])=O)[C@H](O)[C@@H](O)[C@H]1O

InChI

1S/C12H15NO7/c1-6-9(14)10(15)11(16)12(19-6)20-8-4-2-7(3-5-8)13(17)18/h2-6,9-12,14-16H,1H3/t6-,9+,10+,11-,12+/m1/s1

InChI key

YILIDCGSXCGACV-BVWHHUJWSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Nitrophenyl β-D-fucopyranoside has been used as a chromogenic substrate to study substrate specificity and kinetic parameters of β-galactosidase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification, purification and characterization of a novel glycosidase (BgLm1) from Leuconostoc mesenteroides
del Pino-Garcia R, et al.
Food Sci. Technol., 122, 108829-108829 (2020)
New alkalophilic beta-galactosidase with high activity in alkaline pH region from Teratosphaeria acidotherma AIU BGA-1
Yamada M, et al.
Journal of Bioscience and Bioengineering, 123(1), 15-19 (2017)
Purification and analysis of an extremely halophilic beta-galactosidase from Haloferax alicantei
Holmes M L, et al.
Biochimica et Biophysica Acta, Protein Structure and Molecular Enzymology, 1337(2), 276-286 (1997)
Francisco de La Torre et al.
Plant physiology, 128(1), 247-255 (2002-01-15)
An alpha-L-fucosidase (EC 3.2.1.51) able to release the t-fucosyl residue from the side chain of xyloglucan oligosaccharides has been detected in the leaves of Arabidopsis plants. Moreover, an alpha-L-fucosidase with similar substrate specificity was purified from cabbage (Brassica oleracea) leaves
M H Tomassi et al.
Brazilian journal of medical and biological research = Revista brasileira de pesquisas medicas e biologicas, 43(1), 8-12 (2009-12-23)
The manner by which effects of simultaneous mutations combine to change enzymatic activity is not easily predictable because these effects are not always additive in a linear manner. Hence, the characterization of the effects of simultaneous mutations of amino acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service