Skip to Content
Merck
All Photos(6)

Key Documents

N2132

Sigma-Aldrich

4-Nitrophenyl β-D-xylopyranoside

greener alternative

≥98%

Synonym(s):

PNPX

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C11H13NO7
CAS Number:
Molecular Weight:
271.22
Beilstein:
90066
EC Number:
MDL number:
UNSPSC Code:
12352204
PubChem Substance ID:
NACRES:
NA.32

Quality Level

Assay

≥98%

form

powder

greener alternative product score

old score: 100
new score: 51
Find out more about DOZN™ Scoring

greener alternative product characteristics

Waste Prevention
Safer Solvents and Auxiliaries
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

sustainability

Greener Alternative Product

solubility

methanol: 49.00-51.00 mg/mL

greener alternative category

storage temp.

−20°C

SMILES string

O[C@@H]1CO[C@@H](Oc2ccc(cc2)[N+]([O-])=O)[C@H](O)[C@H]1O

InChI

1S/C11H13NO7/c13-8-5-18-11(10(15)9(8)14)19-7-3-1-6(2-4-7)12(16)17/h1-4,8-11,13-15H,5H2/t8-,9+,10-,11+/m1/s1

InChI key

MLJYKRYCCUGBBV-YTWAJWBKSA-N

Looking for similar products? Visit Product Comparison Guide

General description

We are committed to bringing you Greener Alternative Products, which adhere to one of the four categories of Greener Alternatives . This product belongs to category of Re-engineered products, showing key improvements in Green Chemistry Principles “Waste Prevention”, “Safer Solvents and Auxiliaries” and “Enhanced Energy Efficiency”. Click here to view its DOZN scorecard.

Application

4-Nitrophenyl β-D-xylopyranoside has been used as a substrate to assay the activity of β-xylosidase. It has also been used in the pharmaceutical inhibition of heparan sulfate proteoglycan (HSPG) biosynthesis.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Junjarus Sermsathanaswadi et al.
Enzyme and microbial technology, 96, 75-84 (2016-11-23)
A newly isolated endo-β-1,4-xylanase (Xyn10E) from Paenibacillus curdlanolyticus B-6 has a modular structure consisting of a family 22 carbohydrate-binding module (CBM), a glycoside hydrolase (GH) family 10 catalytic domain, two fibronectin type III (Fn3) domains, and a family 3 CBM
Preparation of arabinoxylobiose from rye xylan using family 10 Aspergillus aculeatus endo-1, 4-beta-D-xylanase
Rantanen H, et al.
Carbohydrate Polymers, 68(2), 350-359 (2007)
Hetti Palonen et al.
Applied biochemistry and biotechnology, 117(1), 1-17 (2004-05-06)
The wet oxidation pretreatment (water, oxygen, elevated temperature, and pressure) of softwood (Picea abies) was investigated for enhancing enzymatic hydrolysis. The pretreatment was preliminarily optimized. Six different combinations of reaction time, temperature, and pH were applied, and the compositions of
Beatriz Vieira Dos Santos et al.
Applied biochemistry and biotechnology, 189(1), 37-48 (2019-03-14)
This study evaluated the production of lignocellulose-degrading enzymes by solid-state fermentation (SSF) using a microbial consortium of Aspergillus fumigatus SCBM6 and A. niger SCBM1 (AFN extract). The fungal strains were cultivated in sugarcane bagasse (SCB) and wheat bran (WB) as
T Bravman et al.
FEBS letters, 495(1-2), 115-119 (2001-04-27)
A beta-xylosidase from Bacillus stearothermophilus T-6 was cloned, overexpressed in Escherichia coli and purified to homogeneity. Based on sequence alignment, the enzyme belongs to family 39 glycoside hydrolases, which itself forms part of the wider GH-A clan. The conserved Glu160

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service