
Neostigmine bromide

Synonym(s):
3-(N,N-Dimethylcarbamoyloxy)-N,N,N,-trimethylanilinium bromide, Prostigmine
Empirical Formula (Hill Notation):
C12H19BrN2O2
CAS Number:
Molecular Weight:
303.20
Products
Product Category
Brand
Color
Formula Weight
Gene Alias
Manufacturer
Melting Point (°C)
Physical Form
Purity
Format
Research Area
Quality Segment
Application
API Family
Special Grade
Showing 1-3 of 3