Skip to Content
Merck
All Photos(5)

Key Documents

HPA001341

Sigma-Aldrich

Anti-MECP2 antibody produced in rabbit

enhanced validation

Ab2, Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-MeCP-2 protein antibody produced in rabbit, Anti-MeCP2 antibody produced in rabbit, Anti-Methyl-CpG-binding protein 2 antibody produced in rabbit

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:200- 1:500

immunogen sequence

MVAGMLGLREEKSEDQDLQGLKDKPLKFKKVKKDKKEEKEGKHEPVQPSAHHSAEPAGAGKAETSEGSGSAPAVPEASASPKQRRSIIRDRGPMYDDPTLPEGWTRKLKQRKSGRSAGKY

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MECP2(4204)

Looking for similar products? Visit Product Comparison Guide

Immunogen

methyl CpG binding protein 2

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MECP2 (methyl CpG binding protein 2) gene encodes an X-linked chromosomal protein that binds to methylated DNA. It belongs to the MBD family of proteins having a methyl-CpG binding domain (MBD) that binds to a single methyl-CpG pair. It represses transcription from methylated gene promoters by interacting with histone deacetylase and the corepressor SIN3A. Mutations in this gene cause Rett syndrome, an X-linked dominant neurodevelopmental disorder that is one of the most common causes of mental retardation in females.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74106

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Gevork N Mnatzakanian et al.
Nature genetics, 36(4), 339-341 (2004-03-23)
Rett syndrome is caused by mutations in the gene MECP2 in approximately 80% of affected individuals. We describe a previously unknown MeCP2 isoform. Mutations unique to this isoform and the absence, until now, of identified mutations specific to the previously
Assignment of the gene for methyl-CpG-binding protein 2 (MECP2) to human chromosome band Xq28 by in situ hybridization.
A Vilain et al.
Cytogenetics and cell genetics, 74(4), 293-294 (1996-01-01)
S Kudo
Molecular and cellular biology, 18(9), 5492-5499 (1998-08-26)
Human leukosialin (CD43) is expressed in a cell lineage-specific as well as a differentiation stage-specific fashion. The leukosialin promoter, made up of an Sp1 binding site and a sequence similar to that of an initiator, possesses high transcriptional potential. Previous
M Wan et al.
American journal of human genetics, 65(6), 1520-1529 (1999-12-01)
Rett syndrome (RTT) is a neurodevelopmental disorder characterized by loss of acquired skills after a period of normal development in infant girls. The responsible gene, encoding methyl-CpG binding protein 2 (MeCP2), was recently discovered. Here we explore the spectrum of
Lucile Marion-Poll et al.
The European journal of neuroscience, 39(7), 1234-1244 (2014-04-10)
Long-lasting brain alterations that underlie learning and memory are triggered by synaptic activity. How activity can exert long-lasting effects on neurons is a major question in neuroscience. Signalling pathways from cytoplasm to nucleus and the resulting changes in transcription and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service