Skip to Content
Merck
All Photos(1)

Key Documents

M8445

Sigma-Aldrich

Anti-MLH1 (C-terminal) antibody produced in rabbit

~1 mg/mL, affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-COCA2, Anti-FCC2, Anti-HNPCC, Anti-MGC5172

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
IP
clone:
polyclonal
species reactivity:
human, mouse, rat
citations:
5
technique(s):
immunoprecipitation (IP): 5-10 μg using Jurkat cell lysates

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

antigen 80-85 kDa

species reactivity

human, mouse, rat

packaging

antibody small pack of 25 μL

concentration

~1 mg/mL

technique(s)

immunoprecipitation (IP): 5-10 μg using Jurkat cell lysates

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MLH1(4292)
mouse ... Mlh1(17350)
rat ... Mlh1(81685)

General description

MLH1 is part of a large multi-subunit protein complex of tumor suppressors, DNA damage sensors, and signal transducers, named BRCA1-associated genome surveillance complex (BASC).
MutL homolog 1 (MLH1) is a nucleoprotein and a major component of mismatch repair system. The MLH1 gene is located in chromosome 3p21 and is made up of 19 exons. The protein has a molecular weight of 80kDa.

Specificity

Anti-MLH1 (C-terminal) specifically recognizes human MLH1 (80-85 kDa).

Immunogen

synthetic peptide corresponding to amino acids 591-606 of human MLH1, conjugated to KLH via an N-terminal added cysteine residue. The corresponding peptide sequence is conserved in human, rat, and mouse.

Application

Anti-MLH1 (C-terminal) antibody produced in rabbit has been used in immunoblotting and immunoprecipitation

Biochem/physiol Actions

A hereditary mutation in the MLH1 gene is implicated in nonpolyposis colorectal cancer-2.
MutL homolog 1 (MLH1) has been shown to be involved in promoting colorectal carcinogenesis.

Physical form

Solution in 0.01 M phos­phate buffered saline, pH 7.4, containing 15 mM sodium azide.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, freeze in working aliquots. Repeated freezing and thawing, or storage in “frost-free” freezers, is not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use. Working dilutions should be discarded if not used within 12 hours.

Disclaimer

Unless otherwise stated in our catalog, our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

David W Brammer et al.
Proceedings of the National Academy of Sciences of the United States of America, 115(11), 2806-2811 (2018-03-02)
Over the past two decades, 33 cases of colonic adenocarcinomas have been diagnosed in rhesus macaques (Macaca mulatta) at the nonhuman primate colony of the Keeling Center for Comparative Medicine and Research at The University of Texas MD Anderson Cancer
Y Wang et al.
Genes & development, 14(8), 927-939 (2000-04-27)
We report the identities of the members of a group of proteins that associate with BRCA1 to form a large complex that we have named BASC (BRCA1-associated genome surveillance complex). This complex includes tumor suppressors and DNA damage repair proteins
Haiyan Chen et al.
Journal of cancer research and clinical oncology, 141(12), 2147-2158 (2015-05-20)
As one of the most essential components of mismatch repair system, MutL homolog 1 (MLH1) plays an increasingly implicated role in initiation and promotion of colorectal carcinogenesis, with germ-line mutations in different loci. However, whether a single genetic variant in
Xiaoqing Chen et al.
Nucleic acids research, 41(20), 9325-9338 (2013-08-14)
Exo1-mediated resection of DNA double-strand break ends generates 3' single-stranded DNA overhangs required for homology-based DNA repair and activation of the ATR-dependent checkpoint. Despite its critical importance in inducing the overall DNA damage response, the mechanisms and regulation of the
Shinichiro Fukuhara et al.
Oncotarget, 5(22), 11297-11307 (2014-12-20)
Mismatch repair (MMR) enzymes have been shown to be deficient in prostate cancer (PCa). MMR can influence the regulation of tumor development in various cancers but their role on PCa has not been investigated. The aim of the present study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service