Skip to Content
Products
Cart
0
AE
EN
Products
Products
Applications
Services
Documents
Support
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Biochemicals
Clinical & Diagnostics
Filtration
Greener Alternative Products
Industrial Microbiology
Labware
Materials Science
Molecular Biology & Functional Genomics
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Water Purification
Analytical Chemistry
Cell Culture & Analysis
Chemistry & Synthesis
Clinical & Diagnostics
Environmental & Cannabis Testing
Food & Beverage Testing & Manufacturing
Genomics
Materials Science & Engineering
Microbiological Testing
mAbs Development & Manufacturing
mRNA Development & Manufacturing
Pharma & Biopharma Manufacturing
Protein Biology
Research & Disease Areas
Contract Manufacturing
Contract Testing
Custom Products
Digital Solutions for Life Science
IVD Development & Manufacturing
Product Services
Support
Safety Data Sheets (SDS)
Certificates of Analysis (COA)
Certificates of Origin (COO)
Certificates of Quality (COQ)
Customer Support
Contact Us
Get Site Smart
FAQ
Quality & Regulatory
Calculators & Apps
Webinars
Login
Back
D-(+)-Maltose monohydrate
Synonym(s):
4-
O
-α-D-Glucopyranosyl-D-glucose, Maltobiose
Empirical Formula (Hill Notation):
C
12
H
22
O
11
· H
2
O
CAS Number:
6363-53-7
Molecular Weight:
360.31
EC Number:
200-716-5
Products
Product Category
carbohydrates (3)
disaccharides (2)
Brand
Sigma-Aldrich (3)
Biological Source
Color
Feature
Formula Weight
pH Value
Physical Form
Product Line
Purity
Quality Segment
Application
Optical Isomer
Technique
Filter Products
Showing 1-3 of 3
Compare
Product Number
Description
Pricing
63418
D-(+)-Maltose monohydrate,
BioUltra, ≥99.0% (HPLC)
Expand
View Pricing
M5895
D-(+)-Maltose monohydrate,
≥98% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Expand
View Pricing
M2250
D-(+)-Maltose monohydrate,
Type II, ≥95%
Expand
View Pricing
Choose up to 4 products to compare
Compare
Clear All
English - EN
Learn More