Skip to Content
Merck
All Photos(1)

Key Documents

M5774

Sigma-Aldrich

Anti-Mouse Serum antibody produced in rabbit

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

primary antibodies

clone

polyclonal

contains

15 mM sodium azide

species reactivity

mouse

technique(s)

immunoelectrophoresis: suitable

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Mouse serum comprises hormones, antigens, electrolytes and antibodies. It serves blocking agent in immunoassays.

Specificity

Strong reactivity with normal mouse serum has been determined by immunoelectrophoresis (IEP). This antiserum has not been assayed for interspecies crossreactivity.

Application

Anti-Mouse Serum antibody produced in rabbit has been used in differential fluorescent labelling. It has also been used in differential staining of inner cell mass (ICM) and and trophectoderm (TE) cells. It has been also been used in the removal of zona pellucidae for the isolation of inner cell mass (ICM) from mouse blastocysts.
Anti-Mouse Serum antibody produced in rabbit was used as blocking agent in differential cell count of trophectoderm and inner cell mass cells of mouse embryos.

Physical form

Rabbit Anti-Mouse Serum is provided as a liquid containing 15 mM sodium azide as preservative.

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Raúl Fernández-González et al.
Reproduction (Cambridge, England), 137(2), 271-283 (2008-11-20)
We have reported that in vitro culture (IVC) of preimplantation mouse embryos in the presence of FCS produces long-term effects (LTE) on development, growth and behaviour of the offspring at adult age. To analyse the mechanisms underlying this phenomenon, we
Bi-Ke Zhu et al.
Reproduction, nutrition, development, 44(6), 617-629 (2005-03-15)
The objective of this study was to examine the effect of paternal heat stress on the in vivo development of preimplantation embryos in the mouse. Synchronised B6CBF1 female mice were mated either to a control male mouse or to one
Carla Mulas et al.
Development (Cambridge, England), 146(6) (2019-03-28)
The power of mouse embryonic stem (ES) cells to colonise the developing embryo has revolutionised mammalian developmental genetics and stem cell research. This power is vulnerable, however, to the cell culture environment, deficiencies in which can lead to cellular heterogeneity
Usanee Sanmee et al.
Clinical and experimental reproductive medicine, 43(3), 152-156 (2016-10-01)
We studied the effect of the site of laser zona opening on the complete hatching of mouse blastocysts and the cell numbers of the completely hatched blastocysts. Mouse blastocysts were randomly allocated to the inner cell mass (ICM) group (zona
Effects of partial or complete laser-assisted hatching on the hatching of mouse blastocysts and their cell numbers
Chailert C, et al.
Reproductive Biology and Endocrinology, 11(1), 21-21 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service