Skip to Content
Merck
All Photos(1)

Key Documents

M2634

Sigma-Aldrich

Malic Dehydrogenase from porcine heart

buffered aqueous glycerol solution, 600-1000 units/mg protein (biuret)

Synonym(s):

L-Malate: NAD+ oxidoreductase, MDH, Malate Dehydrogenase

Sign Into View Organizational & Contract Pricing


About This Item

Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54

form

buffered aqueous glycerol solution

specific activity

600-1000 units/mg protein (biuret)

cation traces

NH4+: ≤10 μg/mg protein

foreign activity

Glutamic-Oxalacetic Transaminase ≤0.01%
Glutamic-Pyruvic Transaminase ≤0.01%

shipped in

wet ice

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

General description

Malic dehydrogenase is a mitochondrial isozyme and an important catalyst in the tricarboxylic acid cycle.
Mitochondrial Isozyme

Application

Malic dehydrogenase has been used in a study to assess the comparative molluscicidal action of Ginko biloba extract on snail hosts. It has also been used in a study to investigate the effect of early feed restriction on metabolic programming and compensatory growth in broiler chickens.

Biochem/physiol Actions

Eukaryotic cells contain two different isozymes of malate dehydrogenase: mitochondrial (m-MDH) and soluble or cytoplasmic (s-MDH). This product consists of the mitochondrial form.
The enzyme catalyzes the following reaction:Oxaloacetate + β-NADH → L-Malate + β-NAD
Malic Dehydrogenase can be reversibly inactivated by treatment with pyridoxal 5′-phosphate at 25 °C.

Unit Definition

One unit will convert 1.0 μmole of oxalacetate and β-NADH to L-malate and β-NAD per min at pH 7.5 at 25 °C.

Physical form

Solution in 50% glycerol containing 0.05 M potassium phosphate buffer, pH 7.5

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

X A Zhan et al.
Poultry science, 86(4), 654-660 (2007-03-21)
The effect of early feed restriction on metabolic programming and compensatory growth was studied in broiler chickens. A total of 480 female 1-d-old broiler birds (Aconred) were randomly allocated to ad libitum and feed-restricted groups, each of which was replicated
S S Chen et al.
The Biochemical journal, 151(2), 297-303 (1975-11-01)
1. Pig heart mitochondrial malate dehydrogenase incubated with pyridoxal 5'-phosphate at pH 8.0 and 25 degrees C gradually loses activity. Such inactivation can be largely reversed by dialysis or by addition of L-lysine or L-cysteine, and can be made permanent
Nicholas R Moody et al.
Bio-protocol, 11(24), e4264-e4264 (2022-01-29)
Phosphoenolpyruvate carboxylase (PEPC) catalyzes a critical step in carbon metabolism in plants and bacteria, the irreversible reaction between bicarbonate and phosphoenolpyruvate to produce the C4 compound oxaloacetate. This enzyme is particularly important in the context of C4 photosynthesis, where it
Comparative molluscicidal action of extract of Ginko biloba sarcotesta, arecoline and niclosamide on snail hosts of Schistosoma japonicum
Chen, S., et al.
Pesticide Biochemistry and Physiology, 89, 237-241 (2007)
Tomoko Matsuda et al.
Journal of pharmacological sciences, 112(3), 320-326 (2010-02-23)
We have reported that the differentiation-inducing factors (DIFs) DIF-1 and DIF-3, morphogens secreted from Dictyostelium discoideum, inhibit proliferation of several cancer cells via suppression of the Wnt/beta-catenin signaling pathway. However, the target molecules of DIFs involved in the anti-proliferative effects

Protocols

Spectrophotometric assay evaluates malic dehydrogenase activity using bovine heart enzyme with critical histidine residue at active site.

Spectrophotometric assay evaluates malic dehydrogenase activity using bovine heart enzyme with critical histidine residue at active site.

Spectrophotometric assay evaluates malic dehydrogenase activity using bovine heart enzyme with critical histidine residue at active site.

Spectrophotometric assay evaluates malic dehydrogenase activity using bovine heart enzyme with critical histidine residue at active site.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service