Skip to Content
Merck
All Photos(1)

Key Documents

F1757

Sigma-Aldrich

Fumarase from porcine heart

ammonium sulfate suspension, ≥300 units/mg protein (biuret)

Synonym(s):

L-Malate hydrolyase, Fumarate hydratase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
EC Number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

form

ammonium sulfate suspension

Quality Level

specific activity

≥300 units/mg protein (biuret)

foreign activity

malic dehydrogenase ≤0.1%

storage temp.

2-8°C

General description

Fumarase (FH in human) is a well-known tricarboxylic-acid-cycle enzyme found in both the cytoplasm and mitochondria of all eukaryotes.[1]
Fumarase is mapped to human chromosome 1q43.[2] Fumarase comprises of three domains D1, D2 and D3 and belongs to class II fumarase family.[3]

Application

Fumarase from porcine heart has been used:
  • as a positive control in isocitrate dehydrogenase and α-ketoglutarate dehydrogenase assay[4]
  • for immobilized enzyme based stability assay[5]
  • in fumarate hydratase assays in human lung carcinoma cells (A549)[6]

Fumarase is used as a protein calibration standard in the purification of intact DNA polymerase a/primase from mouse cells.[7]

Biochem/physiol Actions

Fumarase catalyzes the reversible hydration of fumarate to malate. In its mitochondrial form, fumarate is involved in the Krebs Cycle, while the cytosolic form is involved in amino acid metabolism.
Fumarase is essential for regulation of cellular metabolism.[8] Fumarase elicits protective function from double-strand breaks (DSBs) induced by infrared radiation.[9] Loss of function mutations in the fumarase gene is implicated in hereditary leiomyomatosis and renal cell cancer (HLRCC)-syndrome.[2][10]

Unit Definition

One unit will convert 1.0 μmole of L-malate to fumarate per min at pH 7.6 at 25 °C.

Physical form

Suspension in 3.2 M (NH4)2SO4, 0.05 M KH2PO4, pH 7.5, 0.014 M 2-mercaptoethanol

Preparation Note

Isolated by the method of Beeckmans, S., and Kanarek, L., Eur. J. Biochem., 78, 437 (1977). Affinity purified.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Identification of the catalytic mechanism and estimation of kinetic parameters for fumarase
Mescam M, et al.
The Journal of Biological Chemistry, jbc-M110 (2011)
Intact DNA Polymerase a/Primase from Mouse Cells
Journal of Biochemistry, 264, 19407-19415 (1989)
Fumarase Deficiency: A Safe and Potentially Disease Modifying Effect of High Fat/Low Carbohydrate Diet
Ryder B, et al.
JIMD Reports, 40(3), 77?83-77?83 (2018)
Study of an enzyme membrane reactor with immobilized fumarase for production of L-malic acid
Giorno L, et al.
Biotechnology and Bioengineering, 72(1), 77-84 (2001)
Fumarase: a paradigm of dual targeting and dual localized functions
Yogev O, et al.
FEBS Journal, 278(22), 4230-4242 (2011)

Articles

Instructions for working with enzymes supplied as ammonium sulfate suspensions

Protocols

Enzymatic Assay of Fumarase

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service