Skip to Content
Merck
All Photos(2)

Key Documents

M2444

Sigma-Aldrich

Anti-MDC1 antibody, Mouse monoclonal

clone MDC1-50, purified from hybridoma cell culture

Synonym(s):

Anti-Mediator of DNA Damage Checkpoint Protein 1

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified from hybridoma cell culture

antibody product type

primary antibodies

clone

MDC1-50, monoclonal

form

buffered aqueous solution

mol wt

antigen ~250 kDa (2-3 bands)

species reactivity

monkey, human

packaging

antibody small pack of 25 μL

technique(s)

immunocytochemistry: suitable
immunoprecipitation (IP): suitable
microarray: suitable
western blot: 1-2 μg/mL using total cell extract of G361 cells

isotype

IgG2a

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MDC1(9656)

General description

Monoclonal Anti-MDC1 (mouse IgG2a isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells (NS1 cells) and splenocytes from BALB/c mice immunized with a recombinant human MDC1 fragment. Mediator of DNA damage checkpoint protein 1 (MDC1) contains 2089 amino-acid residues with a predicted molecular weight of 226.4 kDa. The protein contains an FHA (forkhead-associated) domain at its amino terminus and two BRCT (BRCA1 carboxyl terminal) domains at its carboxy terminus.

Specificity

Mouse monoclonal clone MDC1-50 anti-MDC1 antibody recognizes human and monkey MDC1.

Immunogen

recombinant human MDC1 fragement (amino acids 2-200).

Application

Monoclonal Anti-MDC1 antibody produced in mouse has been used in:
  • enzyme-linked immunosorbent assay (ELISA
  • immunofluorescence
  • western blotting
  • immunoprecipitation
  • immunocytochemistry

Mouse monoclonal clone MDC1-50 anti-MDC1 antibody is used to tag mediator of DNA damage checkpoint protein 1 for detection and quantitation by Western blotting and immunohistochemical (IHC) techniques such as ELISA, immunoblotting (approx. 250 kDa, 2-3 bands), immunoprecipitation and immunocytochemistry. It is used as a probe to determine the roles of mediator of DNA damage checkpoint protein 1 in DNA repair and genomic stability.

Biochem/physiol Actions

Genomic instability caused by the disruption of mechanisms that regulate cell-cycle checkpoints, DNA repair and apoptosis may lead to the development of cancer. ATM and ATR protein kinases are mediated to DNA damage sites by molecular adapters or mediators proteins such as Histone H2AX, Claspin and BRCTmotif containing molecules such as 53BP1, BRCA1, and MDC1 (mediator of DNA damage checkpoint protein 1). MDC1 interacts with the MRE11 complex (containing MRE11, RAD50 and NBS1 proteins). The MRE11 complex is involved in the detection repair and signaling of DNA damage. Upon ionizing radiation MDC1 is hyperphosphorylated by ATM and localizes to nuclear foci together with the MRE11 complex, phosphorylated H2AX and 53BP1. A radio resistant DNA synthesis (RDS) phenotype in cells is formed by down regulation of MDC1 protein expression by siRNA.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 15 mM sodium azide.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

53BP1 nuclear bodies form around DNA lesions generated by mitotic transmission of chromosomes under replication stress
Lukas C, et al.
Nature Cell Biology, 13(3), 243-243 (2011)
DICER, DROSHA and DNA damage response RNAs are necessary for the secondary recruitment of DNA damage response factors
Francia S, et al.
Journal of Cell Science, 129(7), 1468-1476 (2016)
A dual interaction between the DNA damage response protein MDC1 and the RAG1 subunit of the V (D) J recombinase
Coster G, et al.
The Journal of biological chemistry, 287(43), 36488-36498 (2012)
Zi-Zhen Zhang et al.
World journal of gastroenterology, 19(12), 1968-1974 (2013-04-10)
To investigate the expression deficiency of key molecular markers in the homologous recombination pathway. Expression loss of breast cancer type 1 susceptibility protein (BRCA1), ataxia telangiectasia mutated (ATM), ATM-Rad3-related (ATR), mediator of DNA damage checkpoint protein 1 (MDC1) and meiotic
Ilaria Gritti et al.
The EMBO journal, 41(22), e108040-e108040 (2022-10-11)
The ribonuclease DIS3 is one of the most frequently mutated genes in the hematological cancer multiple myeloma, yet the basis of its tumor suppressor function in this disease remains unclear. Herein, exploiting the TCGA dataset, we found that DIS3 plays

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service