Skip to Content
Merck
All Photos(1)

Key Documents

M137

Sigma-Aldrich

Methysergide maleate salt

solid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C21H27N3O2 · C4H4O4
CAS Number:
Molecular Weight:
469.53
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Pricing and availability is not currently available.

form

solid

color

white to off-white

solubility

DMSO: >10 mg/mL
H2O: 2 mg/mL

originator

Novartis

SMILES string

OC(=O)\C=C/C(O)=O.[H][C@@]12Cc3cn(C)c4cccc(C1=C[C@H](CN2C)C(=O)N[C@@H](CC)CO)c34

InChI

1S/C21H27N3O2.C4H4O4/c1-4-15(12-25)22-21(26)14-8-17-16-6-5-7-18-20(16)13(10-23(18)2)9-19(17)24(3)11-14;5-3(6)1-2-4(7)8/h5-8,10,14-15,19,25H,4,9,11-12H2,1-3H3,(H,22,26);1-2H,(H,5,6)(H,7,8)/b;2-1-/t14-,15+,19-;/m1./s1

InChI key

LWYXFDXUMVEZKS-ZVFOLQIPSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Methysergide maleate salt has been used as a serotonin (5HT) receptor antagonist to evaluate the potential systemic effects of methysergide (METH) and 5-HT infusions on production and milk composition in multiparous Holstein cows.[1] It may also be used as a serotonin receptor blocker to identify the serotonergic receptor type involved in the direct excitation of mitral cells in the main olfactory bulb.[2]

Biochem/physiol Actions

Methysergide maleate, also known as sansert,[3] is a semisynthetic ergot alkaloid ergometrine derivative.[4] Methysergide is a serotonin 1(5-HT1) receptor agonist and a nonselective 5-HT2 and 5-HT7 serotonin receptor antagonist.[1][2] Methysergide maleate is used as a pharmacological agent to treat migraines and other vascular headaches. But, prolonged and uncontrolled use of this drug may cause Leriche′s syndrome, angina pectoris, acute ischemia of the limbs. In addition, gastrointestinal side effects, such as abdominal cramps, nausea, and diarrhea have also been observed in few patients.[5]

Features and Benefits

This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 3 Oral

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S D Silberstein
Cephalalgia : an international journal of headache, 18(7), 421-435 (1998-10-30)
Methysergide is a semisynthetic ergot alkaloid ergometrine derivative, introduced in pharmacotherapy for migraine prophylaxis as a specific serotonin (5HT) receptor antagonist. Methysergide is not just a 5HT2 antagonist, it is also a 5HT1 agonist. Open and controlled studies attest to
Abdominal angina as a complication of methysergide maleate therapy.
J Katz et al.
JAMA, 199(2), 124-125 (1967-01-09)
N H Prins et al.
British journal of pharmacology, 127(6), 1431-1437 (1999-08-24)
This study aimed to characterize for the first time in vitro 5-HT4 receptors in the canine gastrointestinal tract. For this purpose, we used circular muscle strips of the canine isolated rectum. In the presence of methysergide (60 microM), 5-HT induced
Acute ischemia of a limb complicating methysergide maleate therapy.
A L URELES et al.
JAMA, 183, 1041-1042 (1963-03-23)
J A Terrón
British journal of pharmacology, 121(3), 563-571 (1997-06-01)
1. The receptor mediating the long-lasting hypotensive effect of intravenous (i.v.) 5-hydroxytryptamine (5-HT) in the rat was originally classified as 5-HT1-like. Since some pharmacological properties of this receptor are closely similar to those for the cloned 5-ht7 receptor, the present

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service