Skip to Content
Merck
All Photos(3)

Documents

11493027001

Roche

IsoStrip Mouse Monoclonal Antibody Isotyping Kit

sufficient for 10 tests

Synonym(s):

hybridoma subtyping

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
23201100

usage

sufficient for 10 tests

Quality Level

manufacturer/tradename

Roche

storage temp.

2-8°C

General description

IsoStrip Mouse Monoclonal Antibody Isotyping Kit is a simple, three-step kit for the rapid characterization of mouse monoclonal antibodies.

Contents
  • Development Tubes, contain lyophilized latex beads, precoated with anti-mouse-Ig antibodies
  • Isotyping Strips, precoated with subclass- and light-chain-specific anti-mouse-Ig antibodies

Specificity

Anti-mouse antibodies, bound to the latex beads, will react with any mouse monoclonal antibody regardless of its isotype.Goat anti-mouse antibodies, immobilized to the strip, specifically bind to each of the common mouse antibody isotypes (IgG1, IgG2a, IgG2b, IgG3, IgM, and IgA), and to the light chains.

Specificity for all specific antibodies beside IgML at 1.0 μg/ml: ≥ 2
IgM Lambda, at 1.0 μg/ml: ≥ 1

Application

The kit is used for the rapid identification of class, subclass, and light-chain types of mouse monoclonal antibodies (immunoglobulins).

Features and Benefits

  • Reliable: Yields comparable results to the standard ELISA assay
  • Fast: 5 minutes from start to finish
  • Easy: Only 3 steps
  • Economical: No equipment or additional reagents required
  • Highly specific: No crossreactivity with FBS

Packaging

1 kit containing 2 components.

Specifications

Assay time: 5 minutes
Sample material: Hybridoma supernatant, purified mouse monoclonal antibodies, ascites

Preparation Note

Working solution: Dilute Sample
  • Remove the desired number of isotyping strips from the canister. Remove the caps from an equal number of development tubes.
    The tubes may be labeled with a pencil or felt-tipped lab marker for easy identification.
  • Dilute a sample containing the mouse monoclonal antibody in 1% BSA/ phosphate-buffered saline (PBS), pH 7.2 to 7.6.
    Culture supernatant samples should be diluted 1:10 to 1:100. Ascites samples should be diluted 1:20,000. These are recommended dilutions and may vary depending on the concentration of antibody in your sample. In our experience, a monoclonal antibody
    concentration of 0.1 to 1 g/ml of diluted sample gives the best results. 150 ml of this diluted sample will be added to the development tube.

Other Notes

For life science research only. Not for use in diagnostic procedures.

Legal Information

IsoStrip is a trademark of Roche

Kit Components Only

Product No.
Description

  • Development Tubes, containing lyophilized latex beads, precoated with anti-mouse-Ig antibodies

  • Isotyping Strips, precoated with subclass- and light-chain-specific anti-mouse-Ig antibodies

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

does not flash

Flash Point(C)

does not flash


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Julie A Webster et al.
Cancers, 13(12) (2021-07-03)
The current endoscopy and biopsy diagnosis of esophageal adenocarcinoma (EAC) and its premalignant condition Barrett's esophagus (BE) is not cost-effective. To enable EAC screening and patient triaging for endoscopy, we developed a microfluidic lectin immunoassay, the EndoScreen Chip, which allows
Wioleta M Zelek et al.
Frontiers in immunology, 11, 612402-612402 (2021-01-12)
Better understanding of roles of complement in pathology has fuelled an explosion of interest in complement-targeted therapeutics. The C5-blocking monoclonal antibody (mAb) eculizumab, the first of the new wave of complement blocking drugs, was FDA approved for treatment of Paroxysmal
Shirley Lam et al.
mAbs, 7(6), 1178-1194 (2015-08-26)
Chikungunya virus (CHIKV) is a medically important human viral pathogen that causes Chikungunya fever accompanied with debilitating and persistent joint pain. Host-elicited or passively-transferred monoclonal antibodies (mAb) are essential mediators of CHIKV clearance. Therefore, this study aimed to generate and
Wioleta M Zelek et al.
Immunology, 157(4), 283-295 (2019-05-24)
Over the last decade there has been an explosion in complement therapies; one-third of the drugs in the clinic or in development target C5 protein. Eculizumab, a monoclonal antibody (mAb) that binds C5 and blocks its cleavage by the convertase
V Lascano et al.
Oncogenesis, 4, e136-e136 (2015-01-27)
We have previously shown that the tumor necrosis factor family member a proliferation-inducing ligand (APRIL) enhances intestinal tumor growth in various preclinical tumor models. Here, we have investigated whether APRIL serum levels at time of surgery predict survival in a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service