Skip to Content
Merck
All Photos(3)

Documents

C239

Sigma-Aldrich

CNQX disodium salt hydrate

≥98% (HPLC), solid, AMPA/kainate receptor antagonist.

Synonym(s):

6-Cyano-7-nitroquinoxaline-2,3-dione disodium salt hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C9H2N4Na2O4 · xH2O
CAS Number:
Molecular Weight:
276.12 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

product name

CNQX disodium salt hydrate, ≥98% (HPLC), solid

Quality Level

Assay

≥98% (HPLC)

form

solid

storage condition

protect from light

color

orange to red

solubility

H2O: >2 mg/mL (warmed)

SMILES string

O.[Na+].[Na+].[O-]c1nc2cc(C#N)c(cc2nc1[O-])[N+]([O-])=O

InChI

1S/C9H4N4O4.2Na.H2O/c10-3-4-1-5-6(2-7(4)13(16)17)12-9(15)8(14)11-5;;;/h1-2H,(H,11,14)(H,12,15);;;1H2/q;2*+1;/p-2

InChI key

LCDWHGXMFAZHEO-UHFFFAOYSA-L

Application

CNQX disodium salt hydrate has been used as:
  • as a competitive non-NMDA receptor antagonist and competitive AMPA/kainate receptor antagonist in neuronal cultures(110)
  • as a glutamatergic blockers for measuring inhibitory postsynaptic currents in projection neurons(111)
  • as a AMPA glutamate receptor antagonist prefrontal cortex neurons(112)

Biochem/physiol Actions

CNQX(6-cyano-7-nitroquinoxaline-2,3-dioneis) is a potent, competitive AMPA/kainate receptor antagonist. It is a quinoxaline derivative and also an antagonist for non-N-methyl-d-aspartate (non-NMDA) glutamate receptor. CNQX mediates depolarization thalamic reticular nucleus via α-amino-3-hydroxy-5-methylisoxazole-4-propionic acid receptors (AMPARs). It displays micromolar affinity towards AMPA/kainate receptor.
Potent, competitive AMPA/kainate receptor antagonist.

Features and Benefits

This compound is a featured product for Neuroscience research. Click here to discover more featured Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

CNQX Blocks Kainate-induced Corneal Irritation
Ibitokun, Bernadette O and Miller, Kenneth E
investigate opthalmology and visual science, 53(14), 1855-1855 (2012)
Medicinal chemistry of competitive kainate receptor antagonists
Larsen, Ann M and Bunch, Lennart
ACS Chemical Neuroscience, 2(2), 60-74 (2010)
Selective excitatory actions of DNQX and CNQX in rat thalamic neurons
Lee SH, et al.
Journal of Neurophysiology, 103(4), 1728-1734 (2010)
Alessandro Sanzeni et al.
eLife, 9 (2020-07-01)
Many cortical network models use recurrent coupling strong enough to require inhibition for stabilization. Yet it has been experimentally unclear whether inhibition-stabilized network (ISN) models describe cortical function well across areas and states. Here, we test several ISN predictions, including
Cornelia Schöne et al.
The Journal of neuroscience : the official journal of the Society for Neuroscience, 32(36), 12437-12443 (2012-09-08)
Hypothalamic hypocretin/orexin (hcrt/orx) neurons coordinate sleep-wake cycles, reward seeking, and body energy balance. Neurochemical data suggest that hcrt/orx cells contain several transmitters, but what hcrt/orx cells release onto their projection targets is unknown. A major pathway by which hcrt/orx neurons

Articles

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

DISCOVER Bioactive Small Molecules for Neuroscience

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service