Skip to Content
Merck
All Photos(1)

Key Documents

M0443

Sigma-Aldrich

Mirtazapine

≥98% (HPLC)

Synonym(s):

1,2,3,4,10,14b-Hexahydro-2-methylpyrazino[2,1-a]pyrido[2,3-c][2]benzazepine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H19N3
CAS Number:
Molecular Weight:
265.35
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white

solubility

DMSO: 8 mg/mL, clear

SMILES string

CN1CCN2C(C1)c3ccccc3Cc4cccnc24

InChI

1S/C17H19N3/c1-19-9-10-20-16(12-19)15-7-3-2-5-13(15)11-14-6-4-8-18-17(14)20/h2-8,16H,9-12H2,1H3

InChI key

RONZAEMNMFQXRA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Mirtazapine is a noradrenergic and specific serotonergic antidepressant (NaSSA). Mirtazapine agonizes selective adrenergic and serotonergic receptors so that both NE release and 5-HT1A mediated serotonergic signaling are increased.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

M A Raji et al.
The Annals of pharmacotherapy, 35(9), 1024-1027 (2001-09-28)
Depression in patients with Alzheimer disease is a treatable cause of functional decline, caregiver burden, and mortality. It is often associated with severe weight loss, insomnia, and anxiety. These symptoms independently and collaboratively further worsen the prognosis of these vulnerable
B Milne et al.
British journal of pharmacology, 155(8), 1264-1278 (2008-09-23)
Ultra-low doses of opioid receptor antagonists augment spinal morphine antinociception and block the induction of tolerance. Considering the evidence demonstrating functional and physical interactions between the opioid and alpha(2)-adrenoceptors, this study investigated whether ultra-low doses of alpha(2)-adrenoceptor antagonists also influence
S A Anttila et al.
CNS drug reviews, 7(3), 249-264 (2001-10-19)
The novel antidepressant mirtazapine has a dual mode of action. It is a noradrenergic and specific serotonergic antidepressant (NaSSA) that acts by antagonizing the adrenergic alpha2-autoreceptors and alpha2-heteroreceptors as well as by blocking 5-HT2 and 5-HT3 receptors. It enhances, therefore
David E Kemp
Journal of affective disorders, 169 Suppl 1, S34-S44 (2014-12-24)
The most commonly used pharmacologic therapies for bipolar depression are mood stabilizers, atypical antipsychotics, and antidepressants. This paper reviews common side effects associated with these medications and provides recommendations for managing adverse medication effects in clinical practice. Narrative review based
Katarzyna Kamińska et al.
Pharmacological reports : PR, 66(6), 984-990 (2014-12-03)
The aim of our study was to understand the mechanism of clinical efficacy of the combination of an antidepressant and risperidone in drug-resistant depression. We studied the effect of an antidepressant (mirtazapine) and risperidone (atypical antipsychotic), given separately or jointly

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service