Skip to Content
Merck
All Photos(2)

Key Documents

L9795

Sigma-Aldrich

Lactate Oxidase from Aerococcus viridans

lyophilized powder

Synonym(s):

L-lactate monooxygenase, lactate monooxygenase, lactate oxidative decarboxylase, lactate oxygenase, lactic oxidase, lactic oxygenase

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bacterial (Aerococcus viridans)

Quality Level

form

lyophilized powder

mol wt

80 kDa (gel filtration)

storage temp.

−20°C

General description

Lactate oxidase (LOX) is a flavin mononucleotide (FMN)-dependent enzyme and is a member of the α-hydroxy-acid oxidase flavoenzyme family.

Application

Lactate Oxidase from Aerococcus viridans has been used as a:
  • supplement in the culture medium to convert extracellular lactate to pyruvate
  • coating on glass beads for electrochemical detection of lactate from a single cell by a push-pull nozzle system
  • coating on multi-branched gold nanoparticles (GNPs) for the simultaneous colorimetric detection of lactate in saliva by paper-based multiplexed device

Biochem/physiol Actions

Lactate oxidase (LOX) catalyzes the oxidation of lactate to pyruvate and hydrogen peroxide in the presence of an FMN co-factor. The enzyme aids in the determination of L-lactate indirectly by measuring the hydrogen peroxide formed in the reaction.

Unit Definition

One unit is defined as the amount of enzyme which generates 1m mole of Hydrogen Peroxide per minute at 37°C under standard assay conditions.

Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Resp. Sens. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Nicholas X Williams et al.
Nature electronics, 4(4), 261-268 (2022-04-05)
Electronic waste can lead to the accumulation of environmentally and biologically toxic materials and is a growing global concern. Developments in transient electronics-in which devices are designed to disintegrate after use-have focused on increasing the biocompatibility, whereas efforts to develop
Ingar Leiros et al.
Acta crystallographica. Section F, Structural biology and crystallization communications, 62(Pt 12), 1185-1190 (2006-12-05)
The crystal structure of L-lactate oxidase (LOX) from Aerococcus viridans has been determined at 2.1 A resolution. LOX catalyzes the flavin mononucleotide (FMN) dependent oxidation of lactate to pyruvate and hydrogen peroxide. LOX belongs to the alpha-hydroxy-acid oxidase flavoenzyme family;
Lin Zhou et al.
Analytical chemistry, 93(24), 8680-8686 (2021-06-10)
Metabolism of a single cell, even within the same organization, differs from other cells by orders of magnitude. Single-cell analysis provides key information for early diagnosis of cancer as well as drug screening. Any slight change in the microenvironment may
Cyrielle L Bouchez et al.
STAR protocols, 3(2), 101403-101403 (2022-05-24)
Lactate is a central metabolite in energy metabolism and is also involved in cell signaling and epigenetic regulations. Here, we describe an NADH-independent enzymatic assay allowing rapid, selective, and sensitive quantification of L-lactate down to the pmol range. We detail
Melissa Conti Mazza et al.
Journal of neurochemistry, 162(3), 245-261 (2022-06-18)
Human DJ-1 is a cytoprotective protein whose absence causes Parkinson's disease and is also associated with other diseases. DJ-1 has an established role as a redox-regulated protein that defends against oxidative stress and mitochondrial dysfunction. Multiple studies have suggested that

Articles

Professor Rivnay (Northwestern University, USA) discusses using organic mixed conductors as an alternative to efficiently bridge the ionic world of biology with contemporary microelectronics.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service