Skip to Content
Merck
All Photos(2)

Key Documents

L9783

Sigma-Aldrich

Leupeptin hydrochloride

powder, ≥90% (HPLC)

Synonym(s):

Acetyl-Leu-Leu-Arg-al hydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H38N6O4 · HCl
CAS Number:
Molecular Weight:
463.01
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:
NACRES:
NA.77

Product Name

Leupeptin hydrochloride, microbial, ≥90% (HPLC)

biological source

microbial

Quality Level

Assay

≥90% (HPLC)

form

powder

solubility

water: 50 mg/mL, clear, colorless

storage temp.

−20°C

SMILES string

Cl.CC(C)CC(NC(C)=O)C(=O)NC(CC(C)C)C(=O)NC(CCCNC(N)=N)C=O

InChI

1S/C20H38N6O4.ClH/c1-12(2)9-16(24-14(5)28)19(30)26-17(10-13(3)4)18(29)25-15(11-27)7-6-8-23-20(21)22;/h11-13,15-17H,6-10H2,1-5H3,(H,24,28)(H,25,29)(H,26,30)(H4,21,22,23);1H

InChI key

OQQYPQNFLLAILR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM.
Inhibitor of serine and cysteine proteases. Inhibits plasmin, trypsin, papain, calpain, and cathepsin B. Does not inhibit pepsin, cathepsins A and D, thrombin, or α-chymotrypsin. Effective concentration 10-100 μM. There have been numerous studies using leupeptin to protect against hearing loss caused by acoustic overstimulation or the ototoxic antibiotic gentamicin. (Loss of cochlear hair cells is believed to be mediated by calpain.)

Analysis Note

Leupeptin gives multiple peaks on HPLC because of the formation of tautomeric isomers in solution. Majority of contaminating peptide is racemized leupeptin.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Judi Bryant et al.
Thrombosis research, 122(4), 523-532 (2008-05-23)
ADP plays a key role in platelet aggregation which has led to the development of antiplatelet drugs that target the P2Y12 receptor. The aim of this study was to characterize the effects of two novel P2Y12 receptor antagonists, BX 667
Fangyu Wang et al.
STAR protocols, 2(1), 100295-100295 (2021-02-04)
Cells produce two broad classes of extracellular vesicles (EVs), exosomes and microvesicles (MVs). Exosomes are 30-150 nm vesicles derived from multivesicular bodies, while MVs are 200-1,000 nm vesicles that pinch off from plasma membranes. Reliable isolation of EVs is crucial to understand
Pharmacologic inhibition of site 1 protease activity inhibits sterol regulatory element-binding protein processing and reduces lipogenic enzyme gene expression and lipid synthesis in cultured cells and experimental animals.
Hawkins, J.L., et al.
The Journal of Nutrition, 326, 801-808 (2008)
Martijn J C van der Lienden et al.
Journal of cellular biochemistry, 123(5), 893-905 (2022-03-22)
Glucocerebrosidase (GCase), encoded by the GBA gene, degrades the ubiquitous glycosphingolipid glucosylceramide. Inherited GCase deficiency causes Gaucher disease (GD). In addition, carriers of an abnormal GBA allele are at increased risk for Parkinson's disease. GCase undergoes extensive modification of its
Dragana Rajcic et al.
Redox biology, 41, 101879-101879 (2021-02-08)
Non-alcoholic fatty liver disease (NAFLD) is by now the most prevalent liver disease worldwide. The non-proteogenic amino acid l-citrulline (L-Cit) has been shown to protect mice from the development of NAFLD. Here, we aimed to further assess if L-Cit also

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service