L8404
Linoleic acid methyl ester, cis/trans-isomers
Synonym(s):
Linoleic Acid Methyl Ester Isomers
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
synthetic (organic)
Quality Level
form
liquid
functional group
ester
lipid type
omega FAs
shipped in
ambient
storage temp.
−20°C
Related Categories
Packaging
Sealed ampule
Other Notes
Contains methyl esters of each of the following with the approx. weight percentage of each component as indicated.
trans-9,trans-12-Octadecadienoic acid (50%)
cis-9,trans-12-Octadecadienoic acid (20%)
trans-9,cis-12-Octadecadienoic acid (20%)
cis-9,cis-12-Octadecadienoic acid (10%)
These isomers are found as triglycerides in partially hydrogenated vegetable oils and margarines.
trans-9,trans-12-Octadecadienoic acid (50%)
cis-9,trans-12-Octadecadienoic acid (20%)
trans-9,cis-12-Octadecadienoic acid (20%)
cis-9,cis-12-Octadecadienoic acid (10%)
These isomers are found as triglycerides in partially hydrogenated vegetable oils and margarines.
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Journal of the American Oil Chemists' Society, 69, 95-95 (1992)
Lipids, 30(1), 15-21 (1995-01-01)
The fatty acid composition, total trans content (i.e., sum of all the fatty acids which may have one or more trans double bonds) and geometric and positional isomer distribution of unsaturated fatty acids of 198 human milk samples collected in
Animals : an open access journal from MDPI, 11(8) (2021-08-28)
Lipids of different unsaturation degree were added to dairy ewe diet to test the hypothesis that unsaturated oils would modulate milk fatty acid (FA) profile without impairing or even improving feed efficiency. To this aim, we examined milk FA profile
The Journal of dairy research, 84(4), 453-463 (2017-11-21)
It has been well established that milk yield is affected both by milking frequency and due to the removal of residual milk, but the influence of a combination of these factors is unclear. In this study, four mid-lactation cows were
Journal of dairy science, 101(9), 8429-8445 (2018-06-11)
A direct comparative study of dairy cows and goats was performed to characterize the animal performance and milk fatty acid (FA) responses to 2 types of diets that induce milk fat depression in cows as well as a diet that
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service