Skip to Content
Merck
All Photos(3)

Key Documents

L6669

Sigma-Aldrich

LuminoCt® qPCR ReadyMix

For fast probe-based quantitative PCR

Synonym(s):

LuminoCt® qPCR ReadyMix, LuminoCt®

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.55

Quality Level

form

liquid

usage

sufficient for 100 reactions
sufficient for 2000 reactions
sufficient for 500 reactions

feature

dNTPs included
hotstart

technique(s)

qPCR: suitable

color

colorless

input

purified DNA

suitability

suitable for (quantitative PCR)

application(s)

agriculture

compatibility

Bio-Rad MyiQ ( )
Bio-Rad iCycler iQ
Bio-Rad iQ 5
for use with ABI 5700
for use with ABI 7000
for use with ABI 7300
for use with ABI 7500 Fast
for use with ABI 7500
for use with ABI 7700
for use with ABI 7900 Fast
for use with ABI 7900 HT
for use with ABI 7900
for use with ABI StepOne
for use with ABI StepOnePlus
for use with ABI ViiA 7
for use with Bio-Rad CFX384
for use with Bio-Rad CFX96
for use with Bio-Rad MJ Chromo4
for use with Bio-Rad MJ Opticon 2
for use with Bio-Rad MJ Opticon Cepheid SmartCycler
for use with Bio-Rad MJ Opticon
for use with Bio-Rad MiniOpticon
for use with Eppendorf® Mastercycler ep realplex2 s
for use with Eppendorf® Mastercycler ep realplex
for use with Illumina Eco qPCR
for use with Qiagen Corbett Rotor-Gene 3000
for use with Qiagen Corbett Rotor-Gene 6000
for use with Qiagen Corbett Rotor-Gene Q
for use with Roche LightCycler 480
for use with Strategene Mx3000P
for use with Strategene Mx3005P
for use with Strategene Mx4000

detection method

probe-based

shipped in

wet ice

storage temp.

−20°C

General description

Inclusion of JumpStart Taq antibody in the ReadyMix eliminates polymerase activity at ambient temperatures without causing the performance issues associated with chemically modified hot-start Taq. This allows for very rapid activation of the Taq and delivers unparalleled assay sensitivity, while allowing for benchtop reaction setup.

  • Kit is designed to perform probe-based qPCR assays on commonly available real-time instrument platforms.
  • ReadyMix requires the addition of reference dye (provided in kit) when being used on real-time instruments that require ROX passive reference dye for normalization of qPCR assays.
  • Kit is not compatible with qPCR instruments that utilize glass capillary tubes.
LuminoCt® qPCR ReadyMix combines the performance enhancements of our JumpStart Taq antibody for hot start PCR with the convenience of an easy-to-use reaction mixture.

Application

LuminoCt® qPCR ReadyMix has been used in 2-step quantitative reverse transcription polymerase chain reaction (qRT-PCR) assay to quantify the reverse transcription product or cDNA for gene expression analysis. It has also been used in qPCR for quantifying the RNA extracted from rhinoviruses.

Features and Benefits

  • Assay results in as little as 25 minutes
  • Designed to deliver unsurpassed accuracy, precision, and sensitivity
  • Extensive design and validation of ReadyMix chemistry virtually eliminates the need to optimize assay parameters
  • Ideal for rapid, high-throughput 2-step qPCR protocols
  • The master mix allows consistency and reproducibility from one reaction to the next
  • Reduced preparation time and the risk of contamination from multiple pipetting steps
  • Compatible with commonly available real-time instrument platforms for probe-based qPCR assays
  • Require small amplicons (<200 bp) for optimal results
  • Compatible with commercial primer sets, including TaqMan® assays
  • JumpStart Taq DNA polymerase allows room temperature set-up and the hot-start mechanism prevents primer-dimer and non-specific product formation

Packaging

LuminoCt qPCR ReadyMix, Catalog Number L5794, contains optimized concentrations of Tris-HCl, pH 8.3, KCl, dNTPs (dATP, dCTP, dGTP, TTP), stabilizers, MgCl2 and JumpStart Taq DNA Polymerase. Provided as 100, 500 and 2000 reactions (25 μL mix in a 50 μL reaction volume).

100x ROX internal reference dye, Catalog Number R4526. Optional, for use with machines compatible with an internal reference dye, e.g., ABI and Stratagene.

Default reaction volume is 50 μL

100RXN is packaged as 1 X 2.5 mL
500RXN is packaged as 1 X 12.5 mL
2000RXN is packaged as 1 X 50 mL

Other Notes

  • Commercially available 5′ nuclease assays (e.g., TaqMan® Assays) are compatible with LuminoCt ReadyMix.
  • At a minimum, use software to design primers and probes for assays and ensure that qPCR amplicons are <200 bp.
  • Sigma recommends designing primers and probes with Beacon Designer software

To order primers and probes from Sigma, visit www.sigma.com/oligos

Legal Information

Eppendorf is a registered trademark of Eppendorf AG
JumpStart is a trademark of Sigma-Aldrich Co. LLC
LuminoCt is a registered trademark of Merck KGaA, Darmstadt, Germany
ReadyMix is a trademark of Sigma-Aldrich Co. LLC
TaqMan is a registered trademark of Roche Molecular Systems, Inc.

Kit Components Also Available Separately

Product No.
Description
SDS

  • R4526100X ROX internal reference dyeSDS

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 1


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclooxygenase-1 as a potential therapeutic target for seizure suppression: evidences from zebrafish pentylenetetrazole-seizure model
Barbalho P, et al.
Frontiers in Neurology, 7, 200-200 (2016)
Marc J Champigny et al.
Frontiers in plant science, 4, 230-230 (2013-07-13)
DIR1 is a lipid transfer protein (LTP) postulated to complex with and/or chaperone a signal(s) to distant leaves during Systemic Acquired Resistance (SAR) in Arabidopsis. DIR1 was detected in phloem sap-enriched petiole exudates collected from wild-type leaves induced for SAR
J Martínez et al.
Letters in applied microbiology, 50(6), 603-610 (2010-04-22)
To develop a real-time PCR-based strategy for the detection of Paenibacillus larvae vegetative cells and spores to improve the diagnosis and the screening of American foulbrood (AFB), the most harmful pathology of honeybee brood. A real-time PCR that allowed selective
Judit Abdai et al.
Frontiers in psychology, 7, 1772-1772 (2016-11-30)
Social evaluation is a mental process that leverages the preference toward prosocial partners (positivity bias) against the avoidance of antisocial individuals (negativity bias) in a cooperative context. The phenomenon is well-known in humans, and recently comparative investigations looked at the
Dieffenbach, C. and Dveksler, G., (eds)
PCR Primer: A Laboratory Manual (1995)

Articles

Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.

Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.

Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.

Probe-based QPCR offers high sensitivity and specificity with fluorescent-labeled probes and various dye options for multiplex amplification.

Protocols

Standard qPCR protocol with up to four detection probes at specified concentrations, simplifying reaction setup with LuminoCt ReadyMix.

qPCR protocol template with specific detection probes aids in single-target detection using LuminoCt® ReadyMix™.

qPCR protocol template with specific detection probes aids in single-target detection using LuminoCt® ReadyMix™.

Standard qPCR protocol with up to four detection probes at specified concentrations, simplifying reaction setup with LuminoCt ReadyMix.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service