Skip to Content
Merck
All Photos(1)

Key Documents

69785

Sigma-Aldrich

DL-Lactic acid

~90% (T)

Synonym(s):

2-Hydroxypropionic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OH)COOH
CAS Number:
Molecular Weight:
90.08
Beilstein:
1209341
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

~90% (T)

Quality Level

form

liquid

impurities

water, residual

refractive index

n20/D 1.425 (lit.)

bp

122 °C/15 mmHg (lit.)

density

1.209 g/mL at 25 °C (lit.)

functional group

carboxylic acid
hydroxyl

SMILES string

CC(O)C(O)=O

InChI

1S/C3H6O3/c1-2(4)3(5)6/h2,4H,1H3,(H,5,6)

InChI key

JVTAAEKCZFNVCJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

DL-Lactic acid is a racemic mixture that is used for the synthesis of polylactic acid (PLA) by polycondensation reaction. [1]

Application

DL-Lactic acid can be used to synthesize serine racemase (SR) inhibitors.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1C

Supplementary Hazards

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 1

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Luis E Rojas Tovar et al.
Foods (Basel, Switzerland), 10(2) (2021-02-11)
Non Celiac Wheat Sensitivity (NCWS) is an intolerance to wheat products and individuals with NCWS often adhere to a gluten free diet. However, gluten free diets are often associated with a reduced sensory and nutritional quality. Wheat Germ Agglutinin (WGA)
Design, synthesis, and evaluation of novel inhibitors for wild-type human serine racemase.
Takahara S, et al.
Bioorganic & Medicinal Chemistry Letters, 28(3), 441-445 (2018)
Poly (D, L-lactic acid) homopolymers: Synthesis and characterization
Kaitian, Xu and Kozluca
Turkish Journal of Chemistry, 20, 43-53 (1996)
Jule Anna Horlbog et al.
Scientific reports, 8(1), 12931-12931 (2018-08-30)
The aim of this study was to compare the phenotype of 40 strains of L. monocytogenes under food and host relevant stress conditions. The strains were chosen to represent food and clinical isolates and to be equally distributed between the
Marcella Mauro et al.
Toxicology in vitro : an international journal published in association with BIBRA, 59, 275-280 (2019-04-20)
Dermal exposure to Aluminium nanoparticles (AlNPs) can occur in occupationally- and non occupationally exposed- population, due to the use of Al salts-based antiperspirants. No AlNPs transdermal permeation data exists. Our study investigated in vitro the permeation of 30-60 nm Al2O3NPs dispersed

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service