Skip to Content
Merck
All Photos(6)

Key Documents

L4402

Sigma-Aldrich

Lysostaphin from Staphylococcus staphylolyticus

BioUltra, ≥97% (SDS-PAGE), Protein 40-60 % by biuret, ≥2,000 units/mg protein

Synonym(s):

Glycyl-glycine Endopeptidase

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
Enzyme Commission number:
MDL number:
UNSPSC Code:
12352204
NACRES:
NA.54
Pricing and availability is not currently available.

biological source

bacterial (Staphylococcus sp.)

Quality Level

product line

BioUltra

Assay

≥97% (SDS-PAGE)

form

lyophilized powder

specific activity

≥2,000 units/mg protein

mol wt

25 kDa

composition

Protein, 40-60% biuret

concentration

40—60% protein

technique(s)

cell based assay: suitable

suitability

suitable for cell lysis

antibiotic activity spectrum

Gram-positive bacteria

application(s)

diagnostic assay manufacturing

Mode of action

cell wall synthesis | interferes

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

General description

Chemical structure: peptide
Package size based on protein content

Biochem/physiol Actions

Lysostaphin is a zinc endopeptidase with a molecular weight of approximately 25 kDa. Because lysostaphin cleaves the polyglycine cross-links in the peptidoglycan layer of the cell wall of Staphylococcus species it has been found useful for cell lysis and also as a potential anti-microbial therapeutic.
pH Optimum for activity: ~7.5

Unit Definition

One unit will reduce the turbidity (A620) of a suspension of Staphylococcus aureus cells from 0.250 to 0.125 in 10 min at pH 7.5 at 37 °C in a 6.0 ml reaction mixture.

Physical form

Contains potassium phosphate buffer salts and sodium chloride

Preparation Note

Affinity purified

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Resp. Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Johnny Gutierrez et al.
Open forum infectious diseases, 6(4), ofz090-ofz090 (2019-04-27)
Staphylococcus aureus (SA) bacteremia often requires a long treatment duration with antibiotics to prevent relapse due to the ability of SA to establish reservoirs of infection in sites such as heart and bone. These metastatic sites of infection cannot be
Thomas S Lawson et al.
Journal of clinical laboratory analysis, 25(5), 359-365 (2011-09-16)
Aspects of the fluorescence in situ hybridization (FISH) method for the detection of clinically important bacteria, such as Staphylococcus aureus, Staphylococcus epidermidis, and Escherichia coli, were investigated for optimization. Various approaches to optimizing the FISH procedure were taken and different
R Satishkumar et al.
Nanotechnology, 22(50), 505103-505103 (2011-11-24)
The objective of this paper was to study the effect of antibody-directed targeting of S. aureus by comparing the activities of lysostaphin conjugated to biodegradable polylactide nanoparticles (NPs) in the presence and in the absence of co-immobilized anti-S. aureus antibody.
Igor Belyansky et al.
The American surgeon, 77(8), 1025-1031 (2011-09-29)
Mesh and wound infections during hernia repair are predominantly caused by Staphylococcus aureus. Human acellular dermis (HAD) is known to lose its integrity in the face of large bacterial loads. The goal of this study was to determine if lysostaphin
Yuliya Yurko et al.
Surgical innovation, 19(1), 20-26 (2011-07-12)
Naturally occurring antimicrobial peptides are possibly the "next frontier" in infection prevention. Binding them to mesh could reduce the rate of mesh infections. This study identifies an antimicrobial agent capable of significant antibacterial activity when bound to mesh. Lysozyme, human

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service