Skip to Content
Merck
All Photos(2)

Key Documents

L3892

Sigma-Aldrich

Lectin from Triticum vulgaris (wheat)

peroxidase conjugate, lyophilized powder

Synonym(s):

WGA, Wheat germ agglutinin

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32
Pricing and availability is not currently available.

biological source

Triticum vulgaris

Quality Level

conjugate

peroxidase conjugate

Assay

≥85% protein basis (Warburg-Christian)

form

lyophilized powder

potency

<40 μg/mL agglutination activity

peroxidase activity

50-200 units/mg protein

composition

Protein, ~90% modified Warburg-Christian

storage temp.

−20°C

Looking for similar products? Visit Product Comparison Guide

Application

Detects glycoproteins containing β(1→4)-N-acetyl-D-glucosamine when used with appropriate peroxidase substrate
General Western Blot Protocol:
  • Glycoprotein sample size: 500ng
  • Lectin Concentration: 0.1ug/ml

  1. Load samples at 500 ng of glycoprotein per lane
  2. Run 4-20% Bis-Tris SDS page gel
  3. Transfer gel to a PVDF membrane
  4. Block membrane for 1 hr at RT with RIPA buffer (R0278 Sigma)
  5. Incubate HRP lectin at 0.1ug/ml with RIPA buffer for 2 hours at RT
  6. Wash membrane 5 x 5 minutes with 25ml RIPA buffer
  7. Detect using chemiluminescent substrate (CPS1-120)

Biochem/physiol Actions

WGA is not blood group specific but has an affinity for N-acetyl-β-D-glucosaminyl residues and N-acetyl-β-D-glucosamine oligomers. WGA contains no protein-bound carbohydrate.

Packaging

Package size based on protein content.

Unit Definition

One unit will form 1.0 mg of purpurogallin from pyrogallol in 20 sec at pH 6.0 at 20 °C.

Physical form

Contains citrate buffer

Preparation Note

Conjugates are prepared from affinity purified lectin.
Prepared from peroxidase type VI (P8375) using a modification of the method of O′Sullivan,[1] which favors low molecular weight conjugates. Repurified after conjugation by affinity chromatography.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Xinyu Qi et al.
Journal of bone and joint infection, 6(7), 241-253 (2021-07-16)
The high antibiotic tolerance of Staphylococcus aureus biofilms is associated with challenges for treating periprosthetic joint infection. The toxin-antitoxin system, YefM-YoeB, is thought to be a regulator for antibiotic tolerance, but its physiological role is unknown. The objective of this
Hugo Aguilar-Díaz et al.
PLoS neglected tropical diseases, 4(2), e607-e607 (2010-02-20)
Inhibition of encystment can be conceived as a potentially useful mechanism to block the transmission of Entamoeba histolytica under natural conditions. Unfortunately, amoeba encystment has not been achieved in vitro and drugs inhibiting the formation of cysts are not available.
Qian Guo et al.
Journal of bone and mineral research : the official journal of the American Society for Bone and Mineral Research, 36(10), 2065-2080 (2021-06-23)
ATP-citrate lyase (ACLY), generating most of the nucleocytosolic acetyl coenzyme A (acetyl-CoA) for histone acetylation, links cell metabolism to epigenetic regulation. Recent investigations demonstrated that ACLY activated by metabolic reprogramming played an essential role in both M1 and M2 macrophage
Qian Guo et al.
The Journal of biological chemistry, 101775-101775 (2022-03-09)
It's widely accepted that increasing mitochondrial respiration plays a pivotal role during osteoclastogenesis. Mitochondrial pyruvate carrier (MPC) is the key transporter that links glycolysis to mitochondrial respiration but little is known about its role during osteoclastogenesis. Our goal was to
Xiao Lin et al.
Frontiers in cell and developmental biology, 10, 796227-796227 (2022-05-07)
Osteoporosis, characterized by the destruction of bone resorption and bone formation, is a serious disease that endangers human health. Osteoporosis prevention and treatment has become one of the important research contents in the field of medicine. Acacetin, a natural flavonoid

Articles

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

Find the right lectin for your research with our lectin selection guide, organized by lectin source/species, carbohydrate specificity, blood group specificity, and more.

See All

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service