Skip to Content
Merck
All Photos(1)

Documents

L1030

Sigma-Aldrich

Latex beads, amine-modified polystyrene, fluorescent yellow-green

aqueous suspension, 1.0 μm mean particle size

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352116
NACRES:
NA.56

form

aqueous suspension

composition

Solids, 2.5%

technique(s)

cell based assay: suitable

mean particle size

1.0 μm

fluorescence

λex ~470 nm; λem ~505 nm

application(s)

cell analysis

Looking for similar products? Visit Product Comparison Guide

Application

Latex beads, amine-modified polystyrene, fluorescent yellow-green have been used:
  • in the characterization of nano- and microplastics
  • in tendon preparation to label tendon fascicles to enable precise localization of the tissue surface
  • in phagocytosis assay to label spleen cells

Biochem/physiol Actions

Polystyrene latex beads can be used to create latex agglutination systems. It can be used to study the transmission of Mycobacterium leprae, the causative pathogen of leprosy. These beads are also utilized to develop a method for mass screening for both pulmonary and extrapulmonary tuberculosis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Soumya M Turaga et al.
Neuro-oncology, 22(11), 1591-1601 (2020-06-28)
Glioblastoma (GBM) is the most aggressive primary brain tumor and has a dismal prognosis. Previously, we identified that junctional adhesion molecule A (JAM-A), a cell adhesion molecule, is highly elevated in human GBM cancer stem cells and predicts poor patient
James M May et al.
Archives of biochemistry and biophysics, 440(2), 165-172 (2005-08-02)
Since activated macrophages generate potentially deleterious reactive oxygen species, we studied whether ascorbic acid might function as an antioxidant in these cells. Thioglycollate-elicited murine peritoneal macrophages contained about 3 mM ascorbate that was halved by culture in ascorbate-free medium. However
Ivan Gusachenko et al.
Biophysical journal, 102(9), 2220-2229 (2012-07-25)
Collagen is a triple-helical protein that forms various macromolecular organizations in tissues and is responsible for the biomechanical and physical properties of most organs. Second-harmonic generation (SHG) microscopy is a valuable imaging technique to probe collagen fibrillar organization. In this
Amany Ramah et al.
Veterinary immunology and immunopathology, 220, 109991-109991 (2019-12-20)
Tannin is one of the most common phytochemical secondary phenolic metabolites, which is widely distributed in various plant production systems. Dietary intake of tannin can exert different actions on the immune system. The aim of this study was to evaluate
Jing Liu et al.
Fish & shellfish immunology, 82, 136-146 (2018-08-14)
Clam Ruditapes philippinarum is one of the most important commercial aquaculture species in China. The haemocytes play vital roles in internal defense of the calm. In this investigation, classification and immune functions of R. philippinarum haemocytes were identified. The haemocyte

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service