Skip to Content
Merck
All Photos(1)

Documents

L4530

Sigma-Aldrich

Latex beads, carboxylate-modified polystyrene, fluorescent yellow-green

aqueous suspension, 2.0 μm mean particle size

Synonym(s):

Fluorescent Latex Beads

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12162002
NACRES:
NA.25

form

aqueous suspension

composition

Solids, 2.5%

technique(s)

cell based assay: suitable

mean particle size

2.0 μm

fluorescence

λex ~470 nm; λem ~505 nm

application(s)

cell analysis

Looking for similar products? Visit Product Comparison Guide

Application

Carboxylate-modified polystyrene latex beads have been used to collect morphofunctional data about the immune systems of Carabus lefebvrei, a helicophagous Italian endemic ground beetle often used as an indicator of the habitat quality of the Apennines mountain forests. Carboxylate-modified polystyrene latex beads have also been used to inform comparative studies on immunity defense mechanisms of adults and larvae of the coleopteran Cetonischema aeruginosa and to investigate the phagocytic activity of cultured rat thyroid cells.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Moeto Nagai et al.
Micromachines, 11(4) (2020-04-26)
High throughput reconstruction of in vivo cellular environments allows for efficient investigation of cellular functions. If one-side-open multi-channel microdevices are integrated with micropumps, the devices will achieve higher throughput in the manipulation of single cells while maintaining flexibility and open
Gang Tan et al.
Scientific reports, 8(1), 17828-17828 (2018-12-14)
In this study, we explored the effects of particulate matter 2.5 (PM2.5) eye drops on the ocular surface structure and tear function in mice and established a novel animal model for dry eye research. We found that, following treatment with
S L Mueller-Ortiz et al.
Infection and immunity, 69(12), 7501-7511 (2001-11-14)
Mycobacterium tuberculosis and Mycobacterium avium are facultative intracellular pathogens that are able to survive and replicate in mononuclear phagocytes. Human complement component C3 has previously been shown to mediate attachment and phagocytosis of these bacteria by mononuclear phagocytes. In this
Xiujie Li et al.
The American journal of pathology, 170(1), 399-415 (2007-01-04)
We previously reported that truncation of Notch1 (N1) by provirus insertion leads to overexpression of both the intracellular (N1(IC)) and the extracellular (N1(EC)) domains. We produced transgenic (Tg) mice expressing N1(EC) in T cells and in cells of the myeloid
Pierre F Piguet et al.
European journal of haematology, 68(5), 253-261 (2002-07-30)
Anti-platelet antibodies are known to contribute to some types of thrombocytopenia. In this work we investigated anti-platelet antibodies with opposite influence upon activation and kinetics of platelet caspases. A rabbit anti-platelet antibody induced a profound thrombocytopenia, which was associated with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service