Skip to Content
Merck
All Photos(1)

Key Documents

I9010

Sigma-Aldrich

Anti-Human IgG (Fab specific) antibody produced in goat

whole antiserum

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46
Pricing and availability is not currently available.

biological source

goat

conjugate

unconjugated

antibody form

whole antiserum

antibody product type

secondary antibodies

clone

polyclonal

contains

15 mM sodium azide

technique(s)

indirect ELISA: 1:30,000
quantitative precipitin assay: 4.0 mg/mL

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Immunoglobulin G (IgG) belongs to the immunoglobulin family and is a widely expressed serum antibody. Immunoglobulins have two heavy chains and two light chains connected by a disulfide bond. It is a glycoprotein. IgG is usually found as a monomer. IgG is further subdivided into four classes namely, IgG1, IgG2, IgG3, and IgG4. About 70 percent of the total immunoglobulin consists of IgG.

Specificity

Reactivity is observed versus purified human IgA, IgM, Bence Jones Kappa and Bence Jones Lambda myeloma proteins. No reactivity is observed versus Fc fragment of human IgG.

Application

Anti-Human IgG (Fab specific) antibody produced in goat has been used in immunological assays. It has also been used in cellulose substrates as immobilized anti-hIgG to detect human immunoglobulin G (hIgG) from fluid matrices.
Anti-Human IgG (Fab specific) antibody produced in goat was used in capture ELISA at a dilution of 1:10,000.

Biochem/physiol Actions

IgG antibody subtype is the most abundant of serum immunoglobulins of the immune system. It is secreted by B cells and is found in blood and extracellular fluids and provides protection from infections caused by bacteria, fungi and viruses. Maternal IgG is transferred to fetus through the placenta that is vital for immune defense of the neonate against infections.

Physical form

Supplied as a liquid containing 15 mM sodium azide as a preservative.

Preparation Note

delipidized

Storage and Stability

For continuous use, store at 2-8 °C for up to one month. For extended storage, the solution may be frozen in working aliquots. Repeated freezing and thawing is not recommended. Storage in "frost-free" freezers not recommended. If slight turbidity occurs upon prolonged storage, clarify the solution by centrifugation before use.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hannes Orelma et al.
Biomacromolecules, 13(9), 2802-2810 (2012-07-27)
We investigated the adsorption and chemical conjugation of avidin and its deglycosylated form, neutravidin, on films of regenerated and nanofibrillar cellulose. The dynamics and extent of biomolecular attachment were monitored in situ by quartz crystal microbalance microgravimetry and ex situ
Label-free biodetection using capacitive micromachined ultrasonic transducers (CMUTs) and its application for cardiovascular disease diagnostics
Eaimkhong S, et al.
Journal of nanomedicine & nanotechnology, 3(144), 2-2 (2012)
Jane A Potter et al.
Journal of virology, 86(23), 12923-12932 (2012-09-21)
The E2 envelope glycoprotein of hepatitis C virus (HCV) binds to the host entry factor CD81 and is the principal target for neutralizing antibodies (NAbs). Most NAbs recognize hypervariable region 1 on E2, which undergoes frequent mutation, thereby allowing the
Wei Wang et al.
Journal of pharmaceutical sciences, 96(1), 1-26 (2006-09-26)
The number of therapeutic monoclonal antibody in development has increased tremendously over the last several years and this trend continues. At present there are more than 23 approved antibodies on the US market and an estimated 200 or more are
S Hashira et al.
Pediatrics international : official journal of the Japan Pediatric Society, 42(4), 337-342 (2000-09-15)
Maternal immunoglobulin G (IgG), transferred across the placenta to the fetus during intrauterine life, is an important component of the neonatal immunological defence mechanisms against infection. There is controversy with respect to differences in placental transfer of the different IgG

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service