Skip to Content
Merck
All Photos(2)

Documents

A0293

Sigma-Aldrich

Anti-Human IgG (Fab specific)−Peroxidase antibody produced in goat

affinity isolated antibody

Synonym(s):

Anti Human Fab Antibody, Anti Human Fab Antibody - Anti-Human IgG (Fab specific)-Peroxidase antibody produced in goat

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

goat

conjugate

peroxidase conjugate

antibody form

affinity isolated antibody

antibody product type

secondary antibodies

clone

polyclonal

species reactivity

human

technique(s)

direct ELISA: 1:40,000
dot blot: 1:100,000
immunohistochemistry (formalin-fixed, paraffin-embedded sections): 1:200
western blot (chemiluminescent): 1:100,000

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Human IgGs are glycoprotein antibodies that contain two equivalent light chains and a pair of identical heavy chains. IgGs have four distinct isoforms, ranging from IgG1 to IgG4. These antibodies regulate immunological responses to allergy and pathogenic infections. IgGs have also been implicated in complement fixation and autoimmune disorders,.
Antibody is isolated from anti-human IgG antiserum by immunospecific purification to remove essentially all goat serum proteins, including immunoglobulins, which do not specifically bind to the Fab fragment of human IgG. Anti-Human IgG is conjugated to peroxidase and then further purified to remove unconjugated material.

Immunogen

Fab fragment of human IgG

Application

Anti-Human IgG (Fab specific)-Peroxidase antibody produced in goat has been used in:
  • western blot
  • direct enzyme linked immunosorbent assay (ELISA) at 1:40,000 dilution for analysis of yeast cultures genetically modified to produced recombinant Fab fragments.
  • immunohistochemistry
  • dot blot
  • immunohistochemistry (formalin-fixed, paraffin-embedded sections, 1:200)

Anti-Human IgG antibody was used at a 1:10,000 dilution in PBS and incubated for 1 hour at room temperature. Binding in ELISA assays was detected with a O-phenylene-diamine substrate solution (Sigma).
Goat anti-human IgG (Fab specific)-peroxidase antibody can be used for direct ELISA (1:40,000), dot blot and immunohistochemistry (formalin-fixed, paraffin-embedded sections, 1:200) applications.

Physical form

Solution in 0.01 M phosphate buffered saline, pH 7.4, containing 0.05% MIT

Preparation Note

Prepared by the two-step glutaraldehyde method described by Avrameas, S., et al., Scand. J. Immunol., 8, Suppl. 7, 7 (1978).

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

James Duehr et al.
mSphere, 3(1) (2018-02-13)
Recent reports in the scientific literature have suggested that anti-dengue virus (DENV) and anti-West Nile virus (WNV) immunity exacerbates Zika virus (ZIKV) pathogenesis in vitro and in vivo in mouse models. Large populations of immune individuals exist for a related
Chen Li et al.
Acta pharmacologica Sinica, 32(3), 329-337 (2011-02-01)
To transform the human anti-rabies virus glycoprotein (anti-RABVG) single-chain variable fragment (scFv) into a Fab fragment and to analyze its immunological activity. The Fab gene was amplified using overlap PCR and inserted into the vector pComb3XSS. The recombinant vector was
Hanieh Khalili et al.
Scientific reports, 6, 36905-36905 (2016-11-23)
Infliximab is an antibody that neutralizes TNF-α and is used principally by systemic administration to treat many inflammatory disorders. We prepared the antibody mimetic Fab-PEG-Fab (FpFinfliximab) for direct intravitreal injection to assess whether such formulations have biological activity and potential
Kristell Lebozec et al.
mAbs, 9(6), 945-958 (2017-06-10)
Glycoprotein VI is a platelet-specific collagen receptor critical for in vivo formation of arterial thrombosis. It is also considered as an attractive target for the development of anti-thrombotic drugs because blocking glycoprotein (GP)VI inhibits platelet aggregation without inducing detrimental effects
Deng Ning et al.
Journal of biochemistry, 134(6), 813-817 (2004-02-11)
Anti-HBs Fab fragment has considerable potential for use in the prevention and treatment of liver diseases by HBV. Here we established a high-level expression system to directly produce anti-HBs Fab fragment in Pichia pastoris. This was achieved by co-integration of

Articles

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Review the key factors that should figure in your decision to choose a secondary antibody. Learn about species, subclass, isotype, label, and more.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service