Skip to Content
Merck
All Photos(1)

Key Documents

I6137

Sigma-Aldrich

Monoclonal Anti-Bovine IgM antibody produced in mouse

clone BM-23, ascites fluid

Sign Into View Organizational & Contract Pricing


About This Item

MDL number:
UNSPSC Code:
12352203
NACRES:
NA.46

biological source

mouse

conjugate

unconjugated

antibody form

ascites fluid

antibody product type

secondary antibodies

clone

BM-23, monoclonal

contains

15 mM sodium azide

technique(s)

capture ELISA: suitable
immunoprecipitation (IP): suitable
indirect ELISA: 1:4,000
western blot: suitable

isotype

IgG1

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

General description

Bovine IgMs are glycoprotein antibodies that regulate antigen binding during immunological responses. These antibodies have three isotypes, namely, IgMa, IgMb and IgMc. Structural restraints on movement enhance the complement-fixing functions of bovine IgMs . Monoclonal Anti-Bovine IgM antibody detects an epitope on the heavy chain of bovine IgM. The antibody reacts with bovine serum, but does not react with sera from human, baboon, marmoset, gibbon, rhesus, hamster, rabbit, goat, pig, dog, rat, turkey, chicken, or catfish. Furthermore, the product does not bind to native or reduced bovine light chains.
Monoclonal Anti-Bovine IgM (mouse IgG1 isotype) is derived from the hybridoma produced by the fusion of mouse myeloma cells and splenocytes from an immunized mouse. IgM is highly conserved antibody in vertebrates and is expressed early during immune response. It exists as pentamer and is secreted by peritoneal B cells.

Immunogen

Purified bovine IgM

Application

Monoclonal Anti-Bovine IgM antibody is suitable for use in indirect ELISA (1:1,000), western blot, immunoprecipitation and ELISA.

Biochem/physiol Actions

IgM plays a key role in engulfing of the of apoptotic cells. A reduction in serum IgM levels leads to increased autoimmune response and higher risk for infections. IgM displays polyreactive and autoreactive functionality. It plays a key role in tissue homeostasis by mediating clearance of tissue based molecules.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Escherichia coli O157: H7 and other Shiga toxin-producing E. coli in white veal calves
Cristancho L, et al.
Veterinary Microbiology, 126, 200-209 (2008)
Amy S Austin et al.
Journal of immunology (Baltimore, Md. : 1950), 171(3), 1336-1342 (2003-07-23)
IgA is the predominant Ig isotype in mucosal secretions and thus plays a pivotal role in host defense. The mechanisms by which IgA expression is regulated may differ among species and involve multiple pathways. Various cytokines and costimulators have been
S S Saini et al.
Molecular immunology, 38(5), 389-396 (2001-10-31)
The structure of IgM determined from two cDNAs isolated from a Holstein (BLV7G1) and an Angus x Hereford cross-bred (B5D8) cow reveals high sequence similarity both at nucleotide (98.7%) and amino acid (97.9%) level and is closest to sheep (89.4%).
Surinder S Saini et al.
International immunology, 15(7), 845-853 (2003-06-17)
Naturally occurring antibody repertoires of cattle (Bos taurus) include a group of IgMlambda antibodies with exceptionally long complementarity-determining region 3 of the heavy chain (CDR3H) segments, containing multiple Cys residues. These massive CDR3H segments will greatly influence the tertiary and
Emerging functions of natural IgM and its Fc receptor FCMR in immune homeostasis
Wang H, et al.
Frontiers in Immunology, 7, 99-99 (2016)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service