Skip to Content
Merck
All Photos(1)

Documents

PZ0006

Sigma-Aldrich

Exemestane

≥98% (HPLC)

Synonym(s):

6-Methyleneandrosta-1,4-diene-3,17-dione

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O2
CAS Number:
Molecular Weight:
296.40
MDL number:
UNSPSC Code:
51111800
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

optical activity

[α]/D +250 to +300°, c = 1 in methanol

color

white to off-white

solubility

DMSO: ≥20 mg/mL

storage temp.

2-8°C

SMILES string

C[C@]12CC[C@H]3[C@@H](CC(=C)C4=CC(=O)C=C[C@]34C)[C@@H]1CCC2=O

InChI

1S/C20H24O2/c1-12-10-14-15-4-5-18(22)20(15,3)9-7-16(14)19(2)8-6-13(21)11-17(12)19/h6,8,11,14-16H,1,4-5,7,9-10H2,2-3H3/t14-,15-,16-,19+,20-/m0/s1

InChI key

BFYIZQONLCFLEV-DAELLWKTSA-N

Gene Information

human ... CYP19A1(1588)

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

Exemestane is a steroidal antiestrogen and irreversible aromatase inhibitor. Exemestane acts as a false substrate for the aromatase enzyme. Exemestane also prevents the conversion of androgens to estrogens and is used to treat estrogen-dependent breast cancer.

Features and Benefits

This compound is featured on the Nuclear Receptors (Steroids) page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Pictograms

Health hazardEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Willemien van de Water et al.
The oncologist, 18(1), 8-13 (2012-12-25)
For postmenopausal patients with hormone-sensitive breast cancer, outcome is worse with increasing age at diagnosis. The aim of this study was to assess the incidence of breast cancer recurrence (locoregional and distant), and contralateral breast cancer by age at diagnosis.
Nan Soon Wong et al.
Expert opinion on pharmacotherapy, 6(13), 2353-2363 (2005-10-13)
Breast cancer is a major health problem in developed countries. Endocrine therapy is a key component in the management of hormone receptor-positive disease. Although tamoxifen has historically been the gold standard in the first-line management of early and advanced breast
Willemien van de Water et al.
European journal of cancer (Oxford, England : 1990), 49(2), 297-304 (2012-09-08)
Multiple studies suggest better efficacy of chemotherapy in invasive ductal breast carcinomas (IDC) than invasive lobular breast carcinomas (ILC). However, data on efficacy of adjuvant endocrine therapy regimens and histological subtypes are sparse. This study assessed endocrine therapy efficacy in
Anneleen Lintermans et al.
Expert opinion on drug safety, 10(3), 473-487 (2011-03-25)
Hormone-dependent breast cancer can be successfully treated by either blocking the estrogen receptor, as with tamoxifen, or reducing the production of estrogens, as with aromatase inhibitors. Exemestane is a third-generation aromatase inhibitor used in the treatment of estrogen-receptor-positive breast cancer
Michael Gnant et al.
Journal of the National Cancer Institute, 105(9), 654-663 (2013-02-22)
Breast Cancer Trials of Oral Everolimus 2 (BOLERO-2), a phase III study in postmenopausal women with estrogen receptor-positive breast cancer progressing despite nonsteroidal aromatase inhibitor therapy, showed statistically significant benefits with adding everolimus to exemestane. Moreover, in preclinical studies, mammalian

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service