Skip to Content
Merck
All Photos(1)

Key Documents

I0157

Sigma-Aldrich

Ibudilast

≥99% (HPLC), solid

Synonym(s):

2-Methyl-1-[2-(1-methylethyl)pyrazolo[1,5-a]pyridin-3-yl] 1-propanone, 3-Isobutyryl-2-isopropylpyrazolo[1,5-a]pyridine, KC-404

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14 H18 N2 O
CAS Number:
Molecular Weight:
230.31
MDL number:
UNSPSC Code:
41106305
PubChem Substance ID:
NACRES:
NA.77

Assay

≥99% (HPLC)

form

solid

color

white

solubility

DMSO: soluble 28 mg/mL
H2O: soluble 4.5 mg/mL

storage temp.

2-8°C

SMILES string

CC(C)C(=O)c1c(nn2ccccc12)C(C)C

InChI

1S/C14H18N2O/c1-9(2)13-12(14(17)10(3)4)11-7-5-6-8-16(11)15-13/h5-10H,1-4H3

InChI key

ZJVFLBOZORBYFE-UHFFFAOYSA-N

Gene Information

Biochem/physiol Actions

Phosphodiesterase IV (PDE4) inhbitor. Inhibits platelet aggregation. Anti-asthma drug.

Features and Benefits

This compound is a featured product for Cyclic Nucleotide research. Click here to discover more featured Cyclic Nucleotide products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Barkhof et al.
Neurology, 74(13), 1033-1040 (2010-03-05)
Ibudilast is a phosphodiesterase inhibitor influencing inflammation and neurodegeneration in multiple sclerosis (MS). This study evaluated the safety, tolerability, and effects on MRI parameters of 2 different doses of ibudilast in relapsing forms of MS. In this multicenter, double-blind, phase
Masahiro Noguchi et al.
Biological & pharmaceutical bulletin, 32(11), 1924-1927 (2009-11-03)
Ibudilast (3-isobutyryl-2-isopropylpyrazolo[1,5-alpha]pyridine) is clinically used as a cerebral vasodilator in Japan. However, the effects of ibudilast on retinal blood vessels have not been fully examined. The aim of this study, therefore, was to examine the effects of ibudilast on retinal
Hwa Yoon et al.
Biomedical chromatography : BMC, 24(3), 324-328 (2009-07-25)
A simple, accurate, precise and cost effective reversed-phase HPLC method was developed to determine the concentration of ibudilast in human serum. Ibudilast and an internal standard, butyl 4-hydroxybenzoate, were extracted by liquid-liquid extraction with methyl tert-butyl ether. HPLC analysis was
Michelle Kiebala et al.
PloS one, 6(4), e18633-e18633 (2011-04-16)
Human Immunodeficiency Virus-1 (HIV-1)-associated neurocognitive disorders (HAND) occur, in part, due to the inflammatory response to viral proteins, such as the HIV-1 transactivator of transcription (Tat), in the central nervous system (CNS). Given the need for novel adjunctive therapies for
Primary neuroprotection: the Holy Grail of multiple sclerosis therapy.
Robert J Fox
Neurology, 74(13), 1018-1019 (2010-03-05)

Articles

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Cyclic nucleotides like cAMP modulate cell function via PKA activation and ion channels.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service