Skip to Content
Merck
All Photos(3)

Key Documents

C2313

Sigma-Aldrich

Compound 48/80

powder,  hypotensive agent

Synonym(s):

Poly-p-methoxyphenethylmethylamine

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41106300
NACRES:
NA.77

Product Name

Compound 48/80,

form

powder

Quality Level

solubility

H2O: 50 mg/mL

storage temp.

−20°C

Related Categories

General description

Compound 48/80, is the condensation product of N-methyl p-methoxyphenethylamine and formaldehyde.

Application

Compound 48/80 has been used:
  • to stimulate hemocytes for histamine release for measurement in ELISA and flow cytometry
  • to induce mast cell degranulation
  • to induce anaphylactic activity, dermatitis, pruritis, inflammation in mice

Biochem/physiol Actions

Compound 48/80 (C 48/80) is a hypotensive agent, which promotes release of histamine. C 48/80 reduces the phagocytic ability of hemocytes in Styela plicata. C 48/80 promotes degranulation of mast cells. It Inhibits calmodulin, phospholipase C, endoplasmic reticulum (ER) Ca2+ ATPase and activates G proteins.

Preparation Note

Condensation product of N-methyl-p-methoxyphenethylamine with formaldehyde.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Y Koibuchi et al.
European journal of pharmacology, 115(2-3), 163-170 (1985-09-24)
Compound 48/80 and 14C-labeled compound 48/80 were synthesized, and fractionated by thin-layer chromatography into 14 components (A-N) with various histamine releasing activities and different Ca2+ requirements for their actions. The histamine release induced from rat mast cells in vitro by
Differential activation of the PI 3-kinase effectors AKT/PKB and p70 S6 kinase by compound 48/80 is mediated by PKC alpha
Byrne RD, et al.
Cellular Signalling, 19(2), 321-329 (2007)
C Bronner et al.
Biochimica et biophysica acta, 920(3), 301-305 (1987-08-15)
Compound 48/80 inhibited phosphatidylinositol-specific phospholipase C activity from human platelets. Whereas 1 microgram/ml of compound 48/80 slightly stimulated Ca2+-dependent phospholipase A2, higher concentrations led to dose-dependent inhibition of this platelet enzyme. This biphasic effect was confirmed with phospholipases A2 purified
Development of a short-term canine full-thickness skin organ culture method under serum-free conditions
Abramo F, et al.
American Journal of Animal and Veterinary Sciences, 11(2), 61-69 (2016)
Histamine regulates the inflammatory response of the tunicate Styela plicata
Garcia-Garcia E, et al.
Developmental and Comparative Immunology, 46(2), 382-391 (2014)

Articles

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Discover Bioactive Small Molecules for Lipid Signaling Research

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service