Skip to Content
Merck
All Photos(2)

Key Documents

HT90A

Sigma-Aldrich

Gram Stain Kit For Films

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116121
NACRES:
NA.47

form

solution

shelf life

Expiry date on the label.

IVD

for in vitro diagnostic use

application(s)

hematology
histology

storage temp.

room temp

Application

Films are stained for Gram-positive organisms with crystal violet, mordanted in iodine, decolorized and then stained for Gram-negative organisms with safranin O. For delineating Gram-positive and Gram-negative organisms in films.

Kit Components Only

Product No.
Description

  • Crystal Violet Solution (HT90-1) 8 fl oz

  • Decolorizer Solution (HT90-3) 8 fl oz

  • Gram′s Iodine Solution (HT90-2) 8 fl oz

  • Safranin O Solution (HT90-4) 8 fl oz

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Avital Tidhar et al.
PloS one, 4(9), e7023-e7023 (2009-09-18)
Yersinia pestis is the causative agent of plague. Previously we have isolated an attenuated Y. pestis transposon insertion mutant in which the pcm gene was disrupted. In the present study, we investigated the expression and the role of pcm locus
Avital Tidhar et al.
PLoS neglected tropical diseases, 13(6), e0007449-e0007449 (2019-06-07)
We have previously shown that the cell morphogenesis NlpD lipoprotein is essential for virulence of the plague bacteria, Yersinia pestis. To elucidate the role of NlpD in Y. pestis pathogenicity, we conducted a whole-genome comparative transcriptome analysis of the wild-type

Related Content

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

An overview of the science and practice of bacteriology in clinical diagnostics. Learn more about the application of standard and special stains for microscopic analysis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service