Skip to Content
Merck
All Photos(1)

Documents

94635

Sigma-Aldrich

Gram′s safranin solution

for microscopy

Synonym(s):

Safranin T solution

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H19ClN4
CAS Number:
Molecular Weight:
350.84
Beilstein:
3924099
MDL number:
UNSPSC Code:
12171500
PubChem Substance ID:
NACRES:
MA.02

grade

for microscopy

Quality Level

form

liquid

shelf life

limited shelf life, expiry date on the label

composition

ethanol, 10%

technique(s)

microbe id | staining: suitable

color

red to very dark red

refractive index

n20/D 1.339

solubility

water: soluble at 20 °C

density

0.986 g/mL at 20 °C

εmax

4.0 at 532 nm in 50% ethanol

antibiotic activity spectrum

Gram-negative bacteria
Gram-positive bacteria

application(s)

diagnostic assay manufacturing
hematology
histology

storage temp.

room temp

SMILES string

[Cl-].Cc1cc2nc3cc(C)c(N)cc3[n+](-c4ccccc4)c2cc1N

InChI

1S/C20H18N4.ClH/c1-12-8-17-19(10-15(12)21)24(14-6-4-3-5-7-14)20-11-16(22)13(2)9-18(20)23-17;/h3-11H,1-2H3,(H3,21,22);1H

InChI key

OARRHUQTFTUEOS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

Gram’s safranin solution, also known as Safranin T or Basic Red 2, is a ready-to-use staining solution that comprises 10% safranin in ethanol. Safranin is a basic, cationic, azine dye containing a planar, diaminophenazine chromophore, with a phenyl substituent attached to a ring nitrogen. The trimethyl cationic form of safranine is lipophilic, whereas the dimethyl forms are weakly hydrophilic. It is most notably used in bacteriology as a counterstain in Gram staining to differentiate between Gram-positive and Gram-negative bacteria.

Application

Gram’s safranin solution has been used in the following applications:
  • as a non-toxic replacement for crystal violet for the quantification of biofilm formation
  • to develop a hyaluronic acid visualization method
  • to study improved β-lactam susceptibility against biofilm-embedded staphylococcus aureus by 2-aminothiazole

Features and Benefits

  • Ready-to-use solution.
  • Modified and designed in such a way that staining can be carried out in staining cells, on the staining rack, and in automated staining systems.

Principle

Gram staining enables the differentiation of bacteria into Gram-positive and Gram-negative. The mureine structure of the bacterial cell wall is the basis of the color affinity. Bacteria are first stained with crystal violet, and then with iodine solution, which forms a dye-iodine complex. This complex is retained in the multi-layer mureine structure of the cell wall of Gram-positive bacteria after decolorization, imparting a blue-violet color. The cell wall of Gram-negative bacteria comprises a single-layered murein structure that does not retain the dye-iodine complex, allowing the uptake of safranine counterstain, and imparting a pink-red color.

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

114.8 °F

Flash Point(C)

46 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Quantification of biofilm biomass by staining: Non-toxic safranin can replace the popular crystal violet
Ommen, et al.
Journal of Microbiological Methods, 141, 87-89 (2017)
Conn's Biological Stains
Kiernan & Horobin
Conn?s Biological Stains (2002)
Sahra Kirmusaoglu
Clinical laboratory, 66(1) (2020-02-06)
Due to the emergence of methicillin-resistant Staphylococcus aureus (MRSA) producing biofilm, causing recurrent infections worldwide, new therapeutic combinations need to be discovered to prevent resistance and to make treatment available. It was aimed to improve β-lactam susceptibility against ica-dependent biofilm-embedded
Visualization of a hyaluronan network on the surface of silicone-hydrogel materials
Katarzyna A Wygladacz, Daniel J Hook
Clinical Ophthalmology (Auckland, N.Z.) (2016)
Pernille Ommen et al.
Journal of microbiological methods, 141, 87-89 (2017-08-15)
Crystal violet staining is commonly used for quantification of biofilm formation, although it is highly toxic. Here we test safranin as a non-toxic replacement. Safranin staining provided similar results as crystal violet, but with higher reproducibility. We therefore recommend safranin

Articles

Clostridia are relatively large, gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism.

Clostridia are relatively large, gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism.

Clostridia are relatively large, gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism.

Clostridia are relatively large, gram-positive, rod-shaped bacteria that can undergo only anaerobic metabolism.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service