HS1422
Shallow weigh boat
Medium, square white polystyrene, pk of 500 ea
Synonym(s):
weigh boat, weigh dish, weighing dish, weighing pan
All Photos(4)
About This Item
material
square white polystyrene
packaging
pk of 500 ea
manufacturer/tradename
Heathrow Scientific HS1422
L × W × H
(90 mm) × 90 mm × 7 mm
General description
Our flexible polystyrene boats feature smooth surfaces to provide accurate pour-outs with minimal sample loss.
- Shallow boat with a wide, flat bottom to resist tipping
- Ideal for solid product weighing
- Biologically inert and resistant to dilute and weak acids, aqueous solutions, alcohols, and bases.
- Temperature range -10 to 70°C (14 to 158°F)
- Can be used with microwave moisture analyzers
- All are FDA/food-grade compliant
- Pack of 500
- Disposable