Skip to Content
Merck
All Photos(7)

Key Documents

HPA020046

Sigma-Aldrich

Anti-AKAP4 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-A-kinase anchor protein 4, Anti-A-kinase anchor protein 82 kDa, Anti-AKAP 82, Anti-HI, Anti-Major sperm fibrous sheath protein, Anti-PRKA4, Anti-Protein kinase A-anchoring protein 4, Anti-hAKAP82

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
independent
Learn more about Antibody Enhanced Validation

technique(s)

immunohistochemistry: 1:500- 1:1000

immunogen sequence

EIIVIKDTEKKDQSKTEGSVCLFKQAPSDPVSVLNWLLSDLQKYALGFQHALSPSTSTCKHKVGDTEGEYHRASSENCYSVYADQVNIDYLMNRPQNLRLEMTAAKNTNNNQSPSAPPAKPPSTQRAVISPDGECSIDD

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... AKAP4(8852)

General description

A-kinase anchoring protein 4 (AKAP4) is expressed in the testis and in colon, breast, cervical and ovarian cancer cells

Immunogen

A-kinase anchor protein 4 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

A-kinase anchoring protein 4 (AKAP4) has been shown to be linked with tumors and cancers. It acts as a scaffolding protein and associates with protein kinase A (PKA).

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74312

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Shikha Saini et al.
PloS one, 8(2), e57095-e57095 (2013-03-02)
Breast cancer is the second leading cause of cancer related deaths in women worldwide. Reports about the early diagnosis of breast cancer are suggestive of an improved clinical outcome and overall survival rate in cancer patients. Therefore, cancer screening biomarker
Nirmala Jagadish et al.
Journal of experimental & clinical cancer research : CR, 34, 142-142 (2015-11-23)
Colorectal cancer (CRC) ranks third among the estimated cancer cases and cancer related mortalities in the Western world. Early detection and efficient therapy of CRC remains a major health challenge. Therefore, there is a need to identify novel tumor markers
Kuokuo Li et al.
Journal of assisted reproduction and genetics, 39(1), 251-259 (2021-10-18)
Multiple morphological abnormalities in the sperm flagella (MMAF) comprise a severe phenotype of asthenoteratozoospermia with reduced or absent spermatozoa motility. Whereas dozens of candidate pathogenic genes for MMAF have been identified, the genetic cause in a large proportion of patients
Yong Li et al.
Asian journal of andrology, 23(2), 197-204 (2020-10-11)
Oligoasthenoteratozoospermia (OAT) refers to the combination of various sperm abnormalities, including a decreased sperm count, reduced motility, and abnormal sperm morphology. Only a few genetic causes have been shown to be associated with OAT. Herein, we identified a novel homozygous
Mingrong Lv et al.
International journal of biological sciences, 19(2), 393-411 (2023-01-13)
Asthenoteratozoospermia is one of the major factors for male infertility, whereas the causes of large numbers of cases are still unknown. We identified compound heterozygous variants of FSIP2 in three unrelated individuals from a cohort of 105 patients with asthenoteratozoospermia

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service