Skip to Content
Merck
All Photos(1)

Documents

HPA017245

Sigma-Aldrich

Anti-LRP12 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Low-density lipoprotein receptor-related protein 12 precursor, Anti-Suppressor of tumorigenicity protein 7

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:50-1:200

immunogen sequence

FPVCSPNQASVLENLRLAVRSQLGFTSVRLPMAGRSSNIWNRIFNFARSRHSGSLALVSADGDEVVPSQSTSREPERNHTHRSLFSVESDDTDTENERRDMAGASGGVAAPLPQKVPPTTAVEATVGACASSS

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... LRP12(29967)

General description

LRP12 (low density lipoprotein receptor-related protein 12) is a novel putative transmembrane receptor protein located on human chromosome 8, band q22.2-23.1. It is a member of the low-density lipoprotein receptor (LDLR) superfamily. It possesses unique properties of transmembrane receptor. It is widely expressed in normal human tissues and is particularly abundant in human heart and skeletal muscle.

Immunogen

Low-density lipoprotein receptor-related protein 12 precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

LRP12 (low density lipoprotein receptor-related protein 12) is involved in signal transduction pathways and endocytosis. Its several motifs present in cytoplasmic domain participates in endocytosis and signal transduction. It has been suggested that LRP12 may act as an oncogene in oral tumors and perform a significant role in the transformation process of these cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST72057

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cathie Garnis et al.
Oncogene, 23(14), 2582-2586 (2003-12-17)
Chromosome 8q amplification is a common event observed in cancer. In this study, we used high-resolution array comparative genomic hybridization to resolve two neighboring regions on 8q that are both amplified in oral cancer. One region (at 8q24) contains the
J Qing et al.
Oncogene, 18(2), 335-342 (1999-02-02)
Identification and characterization of genes expressed in normal cells and decreased in their malignant counterparts is an important method for detecting candidate tumor suppressors. Using differential display of mRNAs from nontumorigenic infinite life span human fibroblast cell strain MSU-1.1 and
Michele A Battle et al.
Biochemistry, 42(24), 7270-7282 (2003-06-18)
In 1997, McCormick and co-workers identified a novel putative tumor suppressor gene, designated ST7, encoding a unique protein with transmembrane receptor characteristics [Qing et al. (1999) Oncogene 18, 335-342]. Using degenerate primers corresponding to the highly conserved region of the

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service