Skip to Content
Merck
All Photos(6)

Documents

HPA015310

Sigma-Aldrich

Anti-MYH11 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Myosin heavy chain 11, Anti-Myosin heavy chain, smooth muscle isoform, Anti-Myosin-11, Anti-SMMHC

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

VTVLKKALDEETRSHEAQVQEMRQKHAQAVEELTEQLEQFKRAKANLDKNKQTLEKENADLAGELRVLGQAKQEVEHKKKKLEAQVQELQSKCSDGE

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... MYH11(4629)

General description

MYH11 (myosin, heavy chain 11, smooth muscle) forms a part of the vascular smooth muscle cell (VSMC) along with ACTA2. It is a β-myosin heavy chain isoform specific to smooth muscle cells (SMCs). It exists as a multimeric protein, with each monomeric unit composed of two pairs of light chains in association with two heavy chains. This gene is localized to human chromosome 16p13.11.

Immunogen

Myosin-11 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

MYH11 (myosin, heavy chain 11, smooth muscle) is organized into thick filaments, which lie adjacent to thin filaments made of α-actin. Myosin heavy chain generates force, resulting in the thick filaments sliding along with thin filaments, causing the contraction of smooth muscle. Mutations in this gene lead to abnormal SMC contractile function, and are responsible for familial thoracic aortic aneurysms and dissections (TAAD). The levels of MYH11 are increased post percutaneous transluminal coronary angioplasty (PTCA), and MYH11 has potential as a marker to determine the extent of arterial wall injury and to predict restenosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST71238

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Diana G Bohannon et al.
Neurobiology of aging, 96, 128-136 (2020-10-02)
Lax phenotypic characterization of these morphologically distinct pericytes has delayed our understanding of their role in neurological disorders. We herein establish markers which uniquely distinguish different subpopulations of human brain microvascular pericytes and characterize them independently from cerebrovascular smooth muscle
Esmeralda Vásquez-Pacheco et al.
Theranostics, 14(9), 3603-3622 (2024-07-01)
Background: Myofibroblasts (MYFs) are generally considered the principal culprits in excessive extracellular matrix deposition and scar formation in the pathogenesis of lung fibrosis. Lipofibroblasts (LIFs), on the other hand, are defined by their lipid-storing capacity and are predominantly found in
Magdalena Harakalova et al.
European journal of human genetics : EJHG, 21(5), 487-493 (2012-09-13)
Thoracic aortic aneurysms and dissections (TAAD) is a serious condition with high morbidity and mortality. It is estimated that 20% of non-syndromic TAAD cases are inherited in an autosomal-dominant pattern with variable expression and reduced penetrance. Mutations in myosin heavy
Y Tsuchio et al.
Japanese heart journal, 41(2), 131-140 (2000-06-13)
Recently a novel biochemical method that uses an immunoassay to quantitate serum smooth muscle myosin heavy chain (SMMHC) levels was developed for diagnosis of aortic dissection.) The purpose of this study was to determine whether SMMHC released from the coronary
Shao-Qing Kuang et al.
PLoS genetics, 7(6), e1002118-e1002118 (2011-06-24)
Chromosomal deletions or reciprocal duplications of the 16p13.1 region have been implicated in a variety of neuropsychiatric disorders such as autism, schizophrenia, epilepsies, and attention-deficit hyperactivity disorder (ADHD). In this study, we investigated the association of recurrent genomic copy number

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service